Skip to Content
Merck
All Photos(1)

Key Documents

B150

Sigma-Aldrich

BQ-123

≥99%, sodium salt, lyophilized powder

Synonym(s):

BQ123, Cyclo(D-Trp-D-Asp-Pro-D-Val-Leu)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H41N6O7 Na
CAS Number:
Molecular Weight:
632.68
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥99%

form

lyophilized powder

storage temp.

−20°C

SMILES string

[H][C@@]12CCCN1C(=O)[C@@H](CC(O)=O)NC(=O)[C@@H](Cc3c[nH]c4ccccc34)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](NC2=O)C(C)C

InChI

1S/C31H42N6O7/c1-16(2)12-21-27(40)33-22(13-18-15-32-20-9-6-5-8-19(18)20)28(41)35-23(14-25(38)39)31(44)37-11-7-10-24(37)29(42)36-26(17(3)4)30(43)34-21/h5-6,8-9,15-17,21-24,26,32H,7,10-14H2,1-4H3,(H,33,40)(H,34,43)(H,35,41)(H,36,42)(H,38,39)/t21-,22+,23+,24-,26+/m0/s1

InChI key

VYCMAAOURFJIHD-PJNXIOHISA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

cyclo-Asp-Pro-Val-Leu-Trp

Application

BQ-123 has been used as a selective endothelin-A receptor antagonist to study its effects on tumor necrosis factor α (TNFα)-induced human airway smooth muscle cells (HASMCs) proliferation. It is also used as endothelin-A receptor antagonist to study its effects on TNFα-induced granulocyte–macrophage colony-stimulating factor (GM-CSF) expression.

Biochem/physiol Actions

BQ-123 is an endothelin(Et) receptor antagonist and is exquisite for endothelin A (ETA) receptor. It is a cyclic pentapeptide.
Selective ETA endothelin receptor antagonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Knobloch et al.
Thorax, 64(12), 1044-1052 (2009-10-24)
There is an urgent need to inhibit endothelin-1 (ET-1) induced chronic inflammatory processes in early stages of lung diseases in order to prevent untreatable irreversible stages often accompanied by lung fibrosis and pulmonary hypertension. Nothing is known about the airway
M Clozel et al.
Life sciences, 52(9), 825-834 (1993-01-01)
The aim of this study was to evaluate the role of endothelin and endothelin ETA receptor in the early cerebral vasoconstriction following subarachnoid hemorrhage (SAH) in the rat. SAH induced by injection of autologous blood in the cisterna magna reduced
F Cosentino et al.
Journal of cardiovascular pharmacology, 22 Suppl 8, S332-S335 (1993-01-01)
The present study was designed to determine whether an endothelinA (ETA)-receptor antagonist BQ-123 (cyclo[Dtrp, Dasp, pro-D-Val-Leu]) or an ET-converting enzyme inhibitor phosphoramidon may prevent development of cerebral vasospasm after subarachnoid hemorrhage (SAH). A "double hemorrhage" canine model of the disease
T D Warner et al.
British journal of pharmacology, 112(1), 207-213 (1994-05-01)
1. Endothelin-1 binds almost irreversibly to its receptors and causes prolonged vasoconstrictions. Here we have studied the reversal of established responses to ET-1 in vivo and in vitro by BQ-123, an ETA receptor-selective antagonist, and/or PD 145065, an ETA/ETB receptor
Francesca Schinzari et al.
Hypertension (Dallas, Tex. : 1979), 71(1), 185-191 (2017-12-06)
Obese patients have vascular dysfunction related to impaired insulin-stimulated vasodilation and increased endothelin-1-mediated vasoconstriction. In contrast to the harmful vascular actions of angiotensin (Ang) II, the angiotensin-converting enzyme 2 product Ang-(1-7) has shown to exert cardiovascular and metabolic benefits in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service