Skip to Content
Merck
All Photos(1)

Key Documents

E9137

Sigma-Aldrich

Endothelin 3 human, rat

≥97% (HPLC), powder

Synonym(s):

ET-3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C121H168N26O33S4
CAS Number:
Molecular Weight:
2643.04
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32
Pricing and availability is not currently available.

Quality Level

Assay

≥97% (HPLC)

form

powder

composition

Peptide content, ≥60%

color

white

UniProt accession no.

storage temp.

−20°C

SMILES string

[H]N[C@H]1CSSC[C@H](NC(=O)[C@H](Cc2ccc(O)cc2)NC(=O)[C@H](Cc3ccc(O)cc3)NC(=O)[C@@H](NC(=O)[C@@H]4CSSC[C@H](NC(=O)[C@@H](NC1=O)[C@H](C)O)C(=O)N[C@@H](Cc5ccccc5)C(=O)N[C@@H]([C@H](C)O)C(=O)N[C@@H](Cc6ccc(O)cc6)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCC(O)=O)C(=O)N4)C(C)C)C(=O)N[C@@H](Cc7cnc[nH]7)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H]([C@@H](C)CC)C(=O)N[C@@H]([C@@H](C)CC)C(=O)N[C@@H](Cc8c[nH]c9ccccc89)C(O)=O

InChI

1S/C121H168N26O33S4/c1-11-62(7)97(117(175)139-89(121(179)180)49-70-53-126-77-25-17-16-24-75(70)77)145-118(176)98(63(8)12-2)144-112(170)88(52-95(157)158)136-105(163)81(44-60(3)4)131-109(167)86(50-71-54-125-59-127-71)134-113(171)90-56-182-181-55-76(124)101(159)146-99(64(9)148)120(178)142-91-57-183-184-58-92(115(173)143-96(61(5)6)116(174)137-84(48-69-32-38-74(152)39-33-69)107(165)132-82(108(166)141-90)46-67-28-34-72(150)35-29-67)140-104(162)80(40-41-93(153)154)130-102(160)78(26-18-20-42-122)129-110(168)87(51-94(155)156)135-103(161)79(27-19-21-43-123)128-106(164)83(47-68-30-36-73(151)37-31-68)138-119(177)100(65(10)149)147-111(169)85(133-114(91)172)45-66-22-14-13-15-23-66/h13-17,22-25,28-39,53-54,59-65,76,78-92,96-100,126,148-152H,11-12,18-21,26-27,40-52,55-58,122-124H2,1-10H3,(H,125,127)(H,128,164)(H,129,168)(H,130,160)(H,131,167)(H,132,165)(H,133,172)(H,134,171)(H,135,161)(H,136,163)(H,137,174)(H,138,177)(H,139,175)(H,140,162)(H,141,166)(H,142,178)(H,143,173)(H,144,170)(H,145,176)(H,146,159)(H,147,169)(H,153,154)(H,155,156)(H,157,158)(H,179,180)/t62-,63-,64?,65?,76-,78-,79-,80-,81-,82-,83-,84-,85-,86-,87-,88-,89-,90-,91-,92-,96-,97-,98-,99-,100-/m0/s1

InChI key

OQGZWNZGVYLIFX-XKIQDWPYSA-N

Gene Information

human ... EDN3(1908)
rat ... Edn3(366270)

Looking for similar products? Visit Product Comparison Guide

Related Categories

Amino Acid Sequence

Cys-Thr-Cys-Phe-Thr-Tyr-Lys-Asp-Lys-Glu-Cys-Val-Tyr-Tyr-Cys-His-Leu-Asp-Ile-Ile-Trp [Disulfide Bridges: 1-15; 3-11]

General description

Endothelin 3 (ET3) belongs to endothelin peptide family, which includes three members, ET-1, -2 and -3. These are 21-amino acid peptides, which are synthesized as precursors. They are converted to biologically active peptides, after being cleaved by proteases. There are two endothelin receptors called ETRA and ETRB, and ET3 binds to ETRB. It is localized to human intestine and colon.[1]

Application

Endothelin 3 has also been used as a ligand for endothelin receptor type B (EDNRB) in ex vivo enteric NCC (eNCC) migration assays.[2]
Endothelin 3 human, rat has been used for culturing neural tube explant culture,[3] and the pharmacological study of endothelin receptors.[4]

Biochem/physiol Actions

Endothelin 3 (ET3) is a vasoconstrictor that acts in a paracrine and autocrine manner, and in intestinal lamina propria causes the degranulation of mast cells. It results in the release of ions from intestinal epithelial cells. In acute and chronic intestinal inflammation, it is overexpressed. It is also up-regulated in colon carcinoma, where it supports cell proliferation and survival. It might be involved in colon homeostasis, and is essential for the survival and proliferation of goblet cells.[1] In mice, it is essential for the development of pigment cells derived from neural crest (NC).[5] Its expression is de-regulated in metastatic melanoma.[6]
Potent vasoconstrictor from vascular endothelial cells.
Potent vasoconstrictor from vascular endothelial cells; preferred agonist for ETB endothelin receptors. Induces the production of VEGF.

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juliano T Freitas et al.
Pigment cell & melanoma research, 34(6), 1084-1093 (2021-07-22)
Endothelins are cytokines expressed in the microenvironment of several tumors. To identify which stromal cells in the melanoma microenvironment respond to endothelin, we injected murine melanoma cell lines B16F10, YUMM1.7, and YUMMER1.7 in a transgenic mouse that overexpresses endothelin 3
V T Yeung et al.
Journal of neuroscience research, 46(6), 686-696 (1996-12-15)
The effect of endothelin-3 (ET-3) on cyclic GMP (cGMP) responses to C-type natriuretic peptide (CNP) was studied in primary cultures of mouse astrocytes. Attenuation of CNP-stimulated cGMP formation by ET-3 was time-dependent, with maximum inhibition achieved at 30 min of
Xiang-jie An et al.
Journal of Huazhong University of Science and Technology. Medical sciences = Hua zhong ke ji da xue xue bao. Yi xue Ying De wen ban = Huazhong keji daxue xuebao. Yixue Yingdewen ban, 33(4), 581-586 (2013-08-02)
Endothelin-3 (ET-3) is aberrantly expressed in both metastatic melanoma tissues and cultured melanoma cells. Our previous work showed that ET-3 could promote survival of metastatic melanoma cells via its altered expression. In this study, we investigated the mechanisms responsible for
J Desmarets et al.
Biochemical and biophysical research communications, 256(2), 357-360 (1999-03-18)
Pharmacological evidence has suggested that endothelin-3 (ET-3) may act via a novel form of ET receptor that is shared by ETA receptor antagonists but not by ETB receptor selective agonists. This study analyses the properties of interaction of ET-3 with
K J Dunn et al.
Proceedings of the National Academy of Sciences of the United States of America, 97(18), 10050-10055 (2000-08-30)
Wnt1 signaling has been implicated as one factor involved in neural crest-derived melanocyte (NC-M) development. Mice deficient for both Wnt1 and Wnt3a have a marked deficiency in trunk neural crest derivatives including NC-Ms. We have used cell lineage-directed gene targeting

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service