Skip to Content
Merck
All Photos(1)

Key Documents

A9719

Sigma-Aldrich

Arachidonyl-2′-chloroethylamide hydrate

≥97% (HPLC), oil

Synonym(s):

ACEA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H36ClNO · xH2O
CAS Number:
Molecular Weight:
365.98 (anhydrous basis)
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

oil

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

storage condition

desiccated

color

brown-red

solubility

DMSO: soluble
H2O: insoluble

storage temp.

−20°C

SMILES string

CCCCC\C=C/C\C=C/C\C=C/C\C=C/CCCC(=O)NCCCl

InChI

1S/C22H36ClNO/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-22(25)24-21-20-23/h6-7,9-10,12-13,15-16H,2-5,8,11,14,17-21H2,1H3,(H,24,25)/b7-6-,10-9-,13-12-,16-15-

InChI key

SCJNCDSAIRBRIA-DOFZRALJSA-N

Gene Information

Biochem/physiol Actions

Potent and selective neuronal CB1 cannabinoid receptor agonist.

Features and Benefits

This compound is featured on the Cannabinoid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Caution

Photosensitive; store under argon or nitrogen.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yahya I Asiri et al.
Anesthesia and analgesia, 127(2), 548-555 (2017-10-11)
Development of new analgesics is limited by shortcomings of existing preclinical screening assays such as wide variations in response, suitability for a narrow range of analgesics, and propensity to induce tissue damage. Our aim was to determine the feasibility of
Kristina B V Døssing et al.
Genes, 9(7) (2018-07-06)
Somatostatin (SST) analogues are used to control the proliferation and symptoms of neuroendocrine tumors (NETs). MicroRNAs (miRNA) are small non-coding RNAs that modulate posttranscriptional gene expression. We wanted to characterize the miRNAs operating under the control of SST to elucidate
Chris L Baker et al.
British journal of pharmacology, 142(8), 1361-1367 (2004-07-28)
The vasoactive effects of the synthetic cannabinoid (CB) arachidonyl-2-chloroethylamide (ACEA) was tested in the knee joints of urethane-anaesthetised rats. Experiments were also performed to determine whether these vasomotor responses could be blocked by the selective CB(1) receptor antagonists AM251 (N-(piperidin-1-yl)-5-(4-iodophenyl)-1-(2,4-dichlorophenyl)-4-methyl-1H-pyrazole-3-carboxamide)
C J Hillard et al.
The Journal of pharmacology and experimental therapeutics, 289(3), 1427-1433 (1999-05-21)
Two subtypes of the cannabinoid receptor (CB1 and CB2) are expressed in mammalian tissues. Although selective antagonists are available for each of the subtypes, most of the available cannabinoid agonists bind to both CB1 and CB2 with similar affinities. We
Akiko Tsuchida et al.
Scientific reports, 8(1), 7017-7017 (2018-05-08)
GalNAc-disialyl Lc4 (GalNAc-DSLc4) was reported as a novel antigen that associated with malignant features of renal cell cancers (RCCs). To clarify roles of GalNAc-DSLc4 in malignant properties of RCCs, we identified B4GalNAc-T2 as a responsible gene for the synthesis of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service