Skip to Content
Merck
All Photos(1)

Key Documents

A8973

Sigma-Aldrich

2-Arachidonyl glycerol

~10 mg/mL, ≥98% (HPLC)

Synonym(s):

(all-Z)-5,8,11,14-Eicosatetraenoic acid 2-hydroxy-1-(hydroxymethyl)ethyl ester, 2-AG, 2-Arachidonoylglycerol, 2-Monoarachidonoylglycerol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H38O4
CAS Number:
Molecular Weight:
378.55
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

description

9:1 mixture of 2-AG and 1-AG

Quality Level

Assay

≥98% (HPLC)

form

solution

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

concentration

~10 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

[H]C(CO)(OC(CCC/C=C\C/C=C\C/C=C\C/C=C\CCCCC)=O)CO

InChI

1S/C23H38O4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-23(26)27-22(20-24)21-25/h6-7,9-10,12-13,15-16,22,24-25H,2-5,8,11,14,17-21H2,1H3/b7-6-,10-9-,13-12-,16-15-

InChI key

RCRCTBLIHCHWDZ-DOFZRALJSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Endogenous cannabinoid receptor agonist.

Caution

Air sensitive

Physical form

acetonitrile solution

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bradley E Alger
Science's STKE : signal transduction knowledge environment, 2005(309), pe51-pe51 (2005-11-10)
Endocannabinoids are a class of fatty acid derivatives defined by their ability to interact with the specific cannabinoid receptors that were originally identified as the targets of Delta9-tetrahydocannabinol (Delta9-THC), the psychoactive component of cannabis. Endocannabinoids have been implicated in a
M Lee et al.
The Journal of pharmacology and experimental therapeutics, 275(2), 529-536 (1995-11-01)
Anandamide (arachidonylethanolamide), isolated from the porcine brain, and 2-arachidonyl-glycerol (2-Ara-Gl), derived from the canine gut, are two recently identified putative endogenous cannabinoid receptor ligands. Both ligands have been reported to possess binding affinity for cannabinoid receptor subtypes, CB1 and CB2.
Ekaterina A Placzek et al.
Neuropharmacology, 55(7), 1095-1104 (2008-09-02)
Anandamide (AEA) and 2-arachidonyl glycerol (2-AG), endogenous ligands for the CB1 and CB2 cannabinoid receptors, are referred to as endocannabinoids because they mimic the actions of delta9-tetrahydrocannabinol (Delta9-THC), a plant-derived cannabinoid. The processes by which AEA and 2-AG are biosynthesized
M Bari et al.
Mini reviews in medicinal chemistry, 6(3), 257-268 (2006-03-07)
Endocannabinoids are amides, esters and ethers of long chain polyunsaturated fatty acids, which act as new lipidic mediators. Anandamide (N-arachidonoylethanolamine; AEA) and 2-arachidonoylglycerol (2-AG) are the main endogenous agonists of cannabinoid receptors, able to mimic several pharmacological effects of (-)-Delta9-tetrahydrocannabinol
Katarzyna Malenczyk et al.
The Journal of biological chemistry, 288(45), 32685-32699 (2013-10-04)
Endocannabinoid signaling has been implicated in modulating insulin release from β cells of the endocrine pancreas. β Cells express CB1 cannabinoid receptors (CB1Rs), and the enzymatic machinery regulating anandamide and 2-arachidonoylglycerol bioavailability. However, the molecular cascade coupling agonist-induced cannabinoid receptor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service