Skip to Content
Merck
All Photos(3)

Documents

A8381

Sigma-Aldrich

L-Asparagine monohydrate

≥99% (TLC)

Synonym(s):

(S)-(+)-2-Aminosuccinamic acid, (S)-2-Aminosuccinic acid 4-amide, L-Aspartic acid 4-amide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2COCH2CH(NH2)COOH · H2O
CAS Number:
Molecular Weight:
150.13
Beilstein:
1723527
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

L-Asparagine monohydrate, ≥99% (TLC)

biological source

synthetic

Assay

≥99% (TLC)

form

powder

color

white to off-white

mp

233-235 °C (lit.)

application(s)

cell analysis
peptide synthesis

SMILES string

[H]O[H].N[C@@H](CC(N)=O)C(O)=O

InChI

1S/C4H8N2O3.H2O/c5-2(4(8)9)1-3(6)7;/h2H,1,5H2,(H2,6,7)(H,8,9);1H2/t2-;/m0./s1

InChI key

RBMGJIZCEWRQES-DKWTVANSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Asparagine biosynthesis is catalyzed by glutamine-dependent asparagine synthetase in mammalian tissues. Elevated levels of free asparagine is observed in plants facing stress in the form of drought or high salt. It is also present in senescing leaves and germinating seeds.

Application

L-Asparagine monohydrate has been used:
  • as a component of Sauton′s and chelated Sauton′s media for mycobacterial growth
  • to test its effect in the induction of Ca2+ flux in rice roots using aequorin luminescence imaging
  • as a component of fermentation medium for the Blakeslea trispora spores

Biochem/physiol Actions

Asparagine has high nitrogen to carbon ratio and is a key regulator for nitrogen storage and transport. Its thermal degradation in the presence of sugars leads to the acrylamide formation in foods. Asparagine serves as an amino acid exchange factor and is essential for amino acid homeostasis. It favors cancer cell proliferation.
L-Asparagine, a proteinogenic amino acid, is used in biomanufacturing cell culture systems for the production of therapeutic recombinant proteins and monoclonal antibodies and in cell culture based research.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asparagine promotes cancer cell proliferation through use as an amino acid exchange factor
Krall AS, et al.
Nature Communications, 7(1), 11457-11457 (2016)
Heterologous expression and functional analysis of rice GLUTAMATE RECEPTOR-LIKE family indicates its role in glutamate triggered calcium flux in rice roots
Ni J, et al.
Rice (New York, N.Y.), 9(1), 9-9 (2016)
Asparagine Synthesis
The Enzymes, 10, 561-580 (1974)
Oxidative stress response and morphological changes of Blakeslea trispora induced by butylated hydroxytoluene during carotene production
Nanou K and Roukas T
Applied Biochemistry and Biotechnology, 160(8), 2415-2423 (2010)
Analysis of Mycobacterial Protein Secretion
Mehra A and Philips JA
Bio-protocol, 4(12), 11457-11457 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service