Skip to Content
Merck
All Photos(1)

Key Documents

11189

Sigma-Aldrich

L-Aspartic acid

≥99.5% (T), BioUltra

Synonym(s):

(S)-(+)-Aminosuccinic acid, (S)-Aminobutanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
133.10
Beilstein:
1723530
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

Product Name

L-Aspartic acid, BioUltra, ≥99.5% (T)

product line

BioUltra

Quality Level

Assay

≥99.5% (T)

form

powder or crystals

optical activity

[α]20/D +24.7±1°, c = 5% in 5 M HCl

impurities

insoluble matter, passes filter test
≤0.3% foreign amino acids

ign. residue

≤0.05% (as SO4)

loss

≤0.1% loss on drying, 110 °C

color

white

mp

>300 °C (dec.) (lit.)

solubility

1 M HCl: 0.5 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤150 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
NH4+: ≤200 mg/kg
Na: ≤100 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

0.5 M in 1 M HCl

UV absorption

λ: 260 nm Amax: 0.20
λ: 280 nm Amax: 0.10

SMILES string

N[C@@H](CC(O)=O)C(O)=O

InChI

1S/C4H7NO4/c5-2(4(8)9)1-3(6)7/h2H,1,5H2,(H,6,7)(H,8,9)/t2-/m0/s1

InChI key

CKLJMWTZIZZHCS-REOHCLBHSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Aspartic acid (Asp) is a non-essential amino acid. It makes up to 5% of the amino acids in a protein. The GAC and GAU codons encode Asp. This is a chiral molecule and occurs in two enantiomers namely L-Asp and D-Asp.

Application

L-Aspartic acid has been used:
  • as a metabolite to study the enzyme–metabolite interactions in the central metabolism of Escherichia coli by nuclear magnetic resonance (NMR)
  • in transmission electron microscopy
  • as a component of complete media for culturing Yeast strain

Biochem/physiol Actions

L-Aspartic acid is associated with protein biosynthesis (proteinogenic) and neurotransmission
Principal neurotransmitter for fast synaptic excitation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aspartic Acid
Johnson EC
reference module in biomedical sciences (2017)
Peptidyl-Asp Metalloendopeptidase
Handbook of Proteolytic Enzymes, 1 (2012)
Tijs Vandoorne et al.
Nature communications, 10(1), 4147-4147 (2019-09-14)
Energy metabolism has been repeatedly linked to amyotrophic lateral sclerosis (ALS). Yet, motor neuron (MN) metabolism remains poorly studied and it is unknown if ALS MNs differ metabolically from healthy MNs. To address this question, we first performed a metabolic
Paola Monti et al.
Journal of visualized experiments : JoVE, (150) (2019-08-20)
The finding that the well-known mammalian P53 protein can act as a transcription factor (TF) in the yeast S. cerevisiae has allowed for the development of different functional assays to study the impacts of 1) binding site [i.e., response element
Maren Diether et al.
Molecular systems biology, 15(8), e9008-e9008 (2019-08-30)
Metabolite binding to proteins regulates nearly all cellular processes, but our knowledge of these interactions originates primarily from empirical in vitro studies. Here, we report the first systematic study of interactions between water-soluble proteins and polar metabolites in an entire biological

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service