Skip to Content
Merck
All Photos(4)

Key Documents

A4159

Sigma-Aldrich

L-Asparagine

BioReagent, suitable for cell culture, suitable for insect cell culture

Synonym(s):

(S)-2-Aminosuccinic acid 4-amide, L-Aspartic acid 4-amide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCOCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
132.12
Beilstein:
1723527
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.75

product line

BioReagent

Quality Level

Assay

≥98% (HPLC)

form

powder

technique(s)

cell culture | insect: suitable
cell culture | mammalian: suitable

color

white

mp

235 °C (dec.) (lit.)

application(s)

agriculture

SMILES string

N[C@@H](CC(N)=O)C(O)=O

InChI

1S/C4H8N2O3/c5-2(4(8)9)1-3(6)7/h2H,1,5H2,(H2,6,7)(H,8,9)/t2-/m0/s1

InChI key

DCXYFEDJOCDNAF-REOHCLBHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-Asparagine is used in cell culture media and is a component of MEM non-essential amino acids solution. L-Asparagine has been shown to enhance ornithine decarboxylase activity in cultured human colon adenocarcinoma Caco-2 cells and in cultured IEC-6 intestinal epithelial cells. Spore germination in Bacillus subtilis has been increased in the presence of L-asparagine.
An isoxazoline RGD mimic platelet GPIIb/IIIa antagonist has been prepared by chiral synthesis with L-asparagine as a starting material. L-Asparagine has been utilized in the synthesis of 4-azalysine building blocks for application to combinatorial chemistry.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of overexpression of nutrient receptors on germination of spores of Bacillus subtilis
Cabrera-Martinez, R. M., et al.
Journal of Bacteriology, 185, 2457-264 (2003)
Homochiral 4-azalysine building blocks: syntheses and applications in solid-phase chemistry
Chhabra, S. R., et al.
The Journal of Organic Chemistry, 67, 4017-4029 (2002)
The enantiospecific synthesis of an isoxazoline. A RGD mimic platelet GPIIb/IIIa antagonist.
Zhang L. H., et al.
The Journal of Organic Chemistry, 62, 2466-270 (1997)
Increased translation efficiency and antizyme-dependent stabilization of ornithine decarboxylase in amino acidsupplemented human colon adenocarcinoma cells, Caco-2.
Chabanon, H., et al.
The Biochemical Journal, 348 (Pt 2), 401-408 (2000)
Interaction of asparagine and EGF in the regulation of ornithine decarboxylase in IEC-6 cells
Ray, R. M., et al.
The American Journal of Physiology, 276 (3 Pt 1), G773-G780 null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service