Skip to Content
Merck
All Photos(2)

Key Documents

A6306

Sigma-Aldrich

Agarase from Pseudomonas atlantica

lyophilized powder, ≥5,000 units/mg protein (Lowry)

Synonym(s):

β-Agarase, Agarose 4-glycanohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
42040102
NACRES:
NA.26

biological source

bacterial (Pseudomonas atlantica)

Quality Level

form

lyophilized powder

specific activity

≥5,000 units/mg protein (Lowry)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Unit Definition

One unit will produce 1.0 μg of reducing sugar (measured as D-galactose) from agar per min at pH 6.0 at 40 °C.

Physical form

Product is formulated with phosphate buffered salts, bovine serum albumin and Agarase. Total protein content ranges between 10 – 30% w/w.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyeok-Jin Ko et al.
Applied and environmental microbiology, 78(9), 3051-3058 (2012-02-22)
Autotransporters have been employed as the anchoring scaffold for cell surface display by replacing their passenger domains with heterologous proteins to be displayed. We adopted an autotransporter (YfaL) of Escherichia coli for the cell surface display system. The critical regions
Ji-Hwan Seok et al.
Biotechnology letters, 34(6), 1041-1049 (2012-02-09)
α-Agarase hydrolyzes the α-1,3 linkage of agarose yielding agaro-oligosaccharides. It is less well characterized than β-agarase. AgaA gene (2.3 kb ORF), encoding the α-agarase from Thalassomonas JAMB A33, was subcloned into both a constitutive and an inducible expression vector. Both
Michael-Paul Vockenhuber et al.
Microbiology (Reading, England), 158(Pt 2), 424-435 (2011-11-15)
Transcriptional regulation of primary and secondary metabolism is well-studied in Streptomyces coelicolor, a model organism for antibiotic production and cell differentiation. In contrast, little is known about post-transcriptional regulation and the potential functions of small non-coding RNAs (sRNAs) in this
Young Hoon Oh et al.
Journal of microbiology and biotechnology, 21(8), 818-821 (2011-08-31)
An agar-degrading bacterium was isolated from the guts of spiny turban shells. It was identified as a Pseudoalteromonas species and named Pseudoalteromonas sp. JYBCL 1. The viscosity of the inoculated agar medium decreased by more than 60% after 20 h
Yuji Hatada et al.
Marine biotechnology (New York, N.Y.), 13(3), 411-422 (2010-08-06)
A gene of unknown function from the genome of the agar-degrading deep-sea bacterium Microbulbifer thermotolerans JAMB-A94(T) was functionally identified as a ι-carrageenase gene. This gene, designated as cgiA, is located together with two β-agarase genes, agaA and agaO in a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service