Skip to Content
Merck
All Photos(5)

Documents

C1768

Sigma-Aldrich

Cytarabine

≥90% (HPLC), crystalline, DNA replication inhibitor

Synonym(s):

(β-D-Arabinofuranosyl)cytosine, Ara-C, Arabinocytidine, Arabinosylcytosine, Cytarabine, Cytosine arabinoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13N3O5
CAS Number:
Molecular Weight:
243.22
Beilstein:
89175
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

Cytosine β-D-arabinofuranoside, crystalline, ≥90% (HPLC)

Assay

≥90% (HPLC)

form

crystalline

storage temp.

2-8°C

SMILES string

NC1=NC(=O)N(C=C1)[C@@H]2O[C@H](CO)[C@@H](O)[C@@H]2O

InChI

1S/C9H13N3O5/c10-5-1-2-12(9(16)11-5)8-7(15)6(14)4(3-13)17-8/h1-2,4,6-8,13-15H,3H2,(H2,10,11,16)/t4-,6-,7+,8-/m1/s1

InChI key

UHDGCWIWMRVCDJ-CCXZUQQUSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Ara-C incorporates into DNA and inhibits DNA replication by forming cleavage complexes with topoisomerase I resulting in DNA fragmentation; does not inhibit RNA synthesis. Anti-leukemia agent.

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hamed I Ali et al.
Bioorganic & medicinal chemistry, 15(1), 242-256 (2006-10-20)
Novel 2-deoxo-2-phenyl-5-deazaflavins and 2-deoxo-2-phenylflavin-5-oxides were prepared as a new class of antitumor agents and showed significant antitumor activities against NCI-H 460, HCT 116, A 431, CCRF-HSB-2, andKB cell lines. In vivo investigation, 2-deoxo-10-methyl-2-phenyl-5-deazaflavin exhibited the effective antitumor activity against A
Jorge Cortes et al.
The Lancet. Oncology, 14(4), 354-362 (2013-03-05)
Tosedostat is a novel oral aminopeptidase inhibitor with clinical activity in a previous phase 1-2 study in elderly patients with relapsed or refractory acute myeloid leukaemia (AML). We aimed to compare two dosing regimens of tosedostat. In this randomised phase
Sonia G Das et al.
Journal of medicinal chemistry, 54(16), 5937-5948 (2011-07-26)
Multidrug resistance (MDR) in cancer is a phenomenon in which administration of a single chemotherapeutic agent causes cross-resistance of cancer cells to a variety of therapies even with different mechanisms of action. Development of MDR against standard therapies is a
Sergio Amadori et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 31(35), 4424-4430 (2013-10-16)
This randomized trial evaluated the efficacy and toxicity of sequential gemtuzumab ozogamicin (GO) and standard chemotherapy in older patients with newly diagnosed acute myeloid leukemia (AML). Patients (n = 472) age 61 to 75 years were randomly assigned to induction
Alberto Diez-Torrubia et al.
Journal of medicinal chemistry, 53(2), 559-572 (2009-12-17)
Here we explore the applicability of the dipeptidyl peptidase IV (DPPIV/CD26) based prodrug approach to a variety of amine-containing drugs. Efficient procedures have been developed for the synthesis of dipeptide and tetrapeptide amide prodrugs including N-acylation protocols of the exocyclic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service