Skip to Content
Merck
All Photos(3)

Key Documents

A5513

Sigma-Aldrich

N-Acetylprocainamide hydrochloride

≥99% (HPLC), powder

Synonym(s):

N-Acetylnovocainamide hydrochloride, Acecainide hydrochloride, NAPA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(CH3CONH)C6H4CONHCH2CH2N(C2H5)2·HCl
CAS Number:
Molecular Weight:
313.82
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥99% (HPLC)

form

powder

mp

184-186 °C (lit.)

solubility

H2O: 50 mg/mL

storage temp.

−20°C

SMILES string

Cl[H].CCN(CC)CCNC(=O)c1ccc(NC(C)=O)cc1

InChI

1S/C15H23N3O2.ClH/c1-4-18(5-2)11-10-16-15(20)13-6-8-14(9-7-13)17-12(3)19;/h6-9H,4-5,10-11H2,1-3H3,(H,16,20)(H,17,19);1H

InChI key

IYEWBJUCJHKLHD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-Acetylprocainamide hydrochloride may be used:
  • as an internal standard for spiking plasma samples for ultra-high-pressure liquid chromatography coupled with a diode array detector (UHPLC-DAD) analysis
  • to test its relaxant effect on tracheal smooth muscle tissue preparations
  • in preparation of complexes with N-acetyl-L-tyrosine methyl ester and N-acetyl-L-phenylalanine methyl ester for studying intermolecular interactions using nuclear magnetic resonance (NMR) spectroscopy studies

N-Acetylprocainamide hydrochloride is a class III antiarrhythmic compound. N-Acetylprocainamide hydrochloride has been used in a study to determine the disposition of procainamide and N-acetylprocainamide in protein-calorie malnutrition. N-Acetylprocainamide hydrochloride has also been used to study pharmacokinetics of procainamide and N-acetylprocainamide in rats.

Biochem/physiol Actions

N-acetyltransferase II in liver catalyzes the conversion of procainamide to N-acetylprocainamide (NAPA).
Class III antiarrhythmic. Increases the duration of the action potential by decreasing the delayed outward potassium current, slightly decreasing the calcium current, and slightly depressing the inward rectifier potassium current. This is the active metabolite of procainamide that does not induce systemic lupus erythematosus.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Jung et al.
Drug metabolism and disposition: the biological fate of chemicals, 13(3), 359-363 (1985-05-01)
The influence of dietary protein deficiency on the disposition of procainamide (PA) and its major metabolite, N-acetylprocainamide (NAPA) was investigated in male Sprague-Dawley rats fed for 4 weeks on a 23 (control) or a 5% (low) protein diet ad libitum.
B L Kamath et al.
Journal of pharmaceutical sciences, 70(3), 299-302 (1981-03-01)
The pharmacokinetics of distribution and elimination of procainamide and its major metabolite, N-actylprocainamide, were studied in rats. Eight rats were selected randomly, and each received intravenously 14C-labeled procainamide hydrochloride (75 mg/kg) or 14C-labeled N-acetylprocainamide hydrochloride (86 mg/kg) according to a
The evidence for complex formation between N-acetyl-l-tyrosine methyl ester and N-acetylprocainamide hydrochloride using NMR spectroscopy
Janik A, et al.
Structural Chemistry, 20(4), 699-707 (2009)
Anusha Balla et al.
Pharmaceutics, 10(2) (2018-03-31)
A simple, sensitive, and reliable reversed-phase, Ultra-High-Pressure Liquid Chromatography (UHPLC) coupled with a Diode Array Detector (DAD) method for the simultaneous determination of Procainamide (PA) and its major metabolite, N-acetylprocainamide (NAPA), in rat plasma was developed and validated. A simple
Subhash Chandra Ghosh et al.
The Journal of organic chemistry, 77(18), 8007-8015 (2012-08-17)
A practical method for the amidation of aldehydes with economic ammonium chloride or amine hydrochloride salts has been developed for the synthesis of a wide variety of amides by using inexpensive copper sulfate or copper(I) oxide as a catalyst and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service