Skip to Content
Merck
All Photos(1)

Key Documents

A5278

Sigma-Aldrich

Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti Mouse Hrp Sigma

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

peroxidase conjugate

antibody form

IgG fraction of antiserum

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

direct ELISA: 1:10,000

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

IgG antibodies are the smallest antibody isotype found in blood and extracellular fluid. These antibodies are capable of fighting bacterial and viral infection. They also regulate several functions like placental transport, cell surface-receptor binding and complement activation. Anti-mouse IgG (whole molecule)-peroxidase antibody can be used in ELISA (diluted 1: 200 in PBST) . It can also be used in western blotting. Goat anti-mouse IgG-peroxidase antibody reacts specifically with mouse IgA, IgG1, IgG2a, IgG2b, IgG3 and IgM myeloma proteins.

Immunogen

Purified mouse IgG.

Application

Anti-mouse IgG (whole molecule)-peroxidase antibody (diluted 1: 300) can be used as a secondary antibody in immunohistochemistry. It can also be used in Ouchterlony double diffusion and immunoelectrophoresis.
Western blot analysis using HRP conjugated goat anti-mouse IgG as the secondary antibody at a dilution of 1:3000 in BSA/PBS was performed to detect IgG antibodies against outer-membrane vexicle proteins.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4 containing 1% bovine serum albumin with preservative.

Preparation Note

Prepared using the periodate method described by Wilson, M.B., and Nakane, P.K., in Immunofluorescence and Related Staining Techniques, Elsevier/North Holland Biomedical Press, Amsterdam, p215 (1978).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Tojo et al.
Clinical chemistry, 34(3), 539-543 (1988-03-01)
Hybridomas obtained by fusing the spleen cells of BALB/c female mice hyperimmunized with heat-killed yeast-form cells of Candida albicans NIH B-792 strain and a mouse myeloma cell line, P3X63Ag8.653, produced antibodies to beta-1,2-linked oligomannosyl residues in the phosphomannan-protein complex of
Sabine Hombach-Klonisch et al.
Toxicological sciences : an official journal of the Society of Toxicology, 90(2), 519-528 (2006-01-25)
Oviduct epithelial cells are important for the nourishment and survival of ovulated oocytes and early embryos, and they respond to the steroid hormones estrogen and progesterone. Endocrine-disrupting polyhalogenated aromatic hydrocarbons (PHAH) are environmental toxins that act in part through the
Lucila O Fukasawa et al.
Journal of medical microbiology, 52(Pt 2), 121-125 (2003-01-25)
A mAb against the NadA protein from Neisseria meningitidis strain 3006 (serosubtype B : 2b : P1.2 : P5.2,8) demonstrated strong bactericidal activity against Brazilian epidemic serogroup B strain N44/89 (B : 4,7 : P1.19,15 : P5.5,7) and a serogroup
Yogikala Prabhu et al.
Endocrinology, 155(7), 2391-2401 (2014-05-16)
Mutations in the PCSK1 gene encoding prohormone convertase 1/3 (PC1/3) are strongly associated with obesity in humans. The PC1/3(N222D) mutant mouse thus far represents the only mouse model that mimics the PC1/3 obesity phenotype in humans. The present investigation addresses
Fabian M Arnold et al.
Scientific reports, 8(1), 9539-9539 (2018-06-24)
Molecular research on mycobacteria relies on a multitude of tools for the genetic manipulation of these clinically important bacteria. However, a uniform set of vectors allowing for standardized cloning procedures is not available. Here, we developed a versatile series of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service