Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352203
NACRES:
NA.46
eCl@ss:
32160702
Conjugate:
peroxidase conjugate
Clone:
polyclonal
Application:
ELISA, IHC, WB
Citations:
318
biological source
goat
Quality Level
conjugate
peroxidase conjugate
antibody form
purified immunoglobulin
antibody product type
secondary antibodies
clone
polyclonal
species reactivity
mouse
manufacturer/tradename
Upstate®
technique(s)
ELISA: suitable, immunohistochemistry: suitable, western blot: suitable
shipped in
dry ice
target post-translational modification
unmodified
Related Categories
General description
Immunoglobulin G (IgG), is one of the most abundant proteins in human serum with normal levels between 8-17 mg/mL in adult blood. IgG is important for our defense against microorganisms and the molecules are produced by B lymphocytes as a part of our adaptive immune response. The IgG molecule has two separate functions; to bind to the pathogen that elicited the response and to recruit other cells and molecules to destroy the antigen. The variability of the IgG pool is generated by somatic recombination and the number of specificities in an individual at a given time point is estimated to be 1011 variants.
Immunogen
Highly purified whole mouse IgG.
Application
Additional Research Applications
ELISA and Immunohistochemistry:
A working dilution of 1:10,000 to 1:40,000 is suggested for ELISA and 1:500 to 1:2,000 for immunohistochemistry.
Optimal antibody dilution for other applications should be determined by the researcher.
ELISA and Immunohistochemistry:
A working dilution of 1:10,000 to 1:40,000 is suggested for ELISA and 1:500 to 1:2,000 for immunohistochemistry.
Optimal antibody dilution for other applications should be determined by the researcher.
Goat Anti-Mouse IgG Antibody, HRP conjugate is an antibody against Mouse IgG for use in ELISA, IH & WB.
Research Category
Secondary & Control Antibodies
Secondary & Control Antibodies
Research Sub Category
Whole Immunoglobulin Secondary Antibodies
Whole Immunoglobulin Secondary Antibodies
Biochem/physiol Actions
Recognizes mouse IgG, both heavy and light chains.
Physical form
Purified goat polyclonal IgG in buffer containing conjugated to horseradish peroxidase lyophilized from 0.02 M Potassium Phosphate, 0.15 M NaCl, pH 7.2, 10 mg/mL BSA, and 0.01% gentamicin sulfate.
Rehydration: Add 500 µL sterile, distilled water containing 50% glycerol, to make a 1 mg/mL stock solution.
Rehydration: Add 500 µL sterile, distilled water containing 50% glycerol, to make a 1 mg/mL stock solution.
Preparation Note
Stable for 1 year at -20ºC from date of receipt.
Analysis Note
Western Blot Analysis:
Suitable for western blotting (dot blot) at 1:1,000 to 1:4,000 dilution.
Suitable for western blotting (dot blot) at 1:1,000 to 1:4,000 dilution.
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class
13 - Non Combustible Solids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Research on promoting periodontal regeneration with human basic fibroblast growth factor-modified bone marrow mesenchymal stromal cell gene therapy.
Zhen Tan, Qing Zhao, Ping Gong, Yang Wu, Na Wei, Quan Yuan, Chuan Wang, Dapeng Liao, Hua Tang
Cytotherapy null
The relationship between transcription initiation RNAs and CCCTC-binding factor (CTCF) localization.
Ryan J Taft et al.
Epigenetics & chromatin, 4, 13-13 (2011-08-05)
Transcription initiation RNAs (tiRNAs) are nuclear localized 18 nucleotide RNAs derived from sequences immediately downstream of RNA polymerase II (RNAPII) transcription start sites. Previous reports have shown that tiRNAs are intimately correlated with gene expression, RNA polymerase II binding and
Swan Lin et al.
Pharmaceutical research, 33(1), 72-82 (2015-08-02)
To gain knowledge of lung clearance mechanisms of inhaled tissue plasminogen activator (tPA). Using an in vivo mouse model and ex vivo murine whole organ cell suspensions, we examined the capability of the lungs to utilize LRP1 receptor-mediated endocytosis (RME)
Tiffany Weinkopff et al.
PloS one, 6(8), e22282-e22282 (2011-08-11)
Lymphangiectasia (dilation of the lymphatic vessel (LV)) is pathognomonic for lymphatic filariasis. In both infected humans and animal models of infection, lymphangiectasia is not restricted to the site of the worm nest, but is found along the infected vessel. These
Jorid T Stuenaes et al.
British journal of pharmacology, 160(1), 116-129 (2010-04-24)
Genetic approaches have documented protein kinase B (PKB) as a pivotal regulator of heart function. Insulin strongly activates PKB, whereas adrenaline is not considered a major physiological regulator of PKB in heart. In skeletal muscles, however, adrenaline potentiates insulin-stimulated PKB
Related Content
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service