Skip to Content
Merck
All Photos(4)

Key Documents

A4034

Sigma-Aldrich

(+)-Sodium L-ascorbate

powder, BioReagent, suitable for cell culture

Synonym(s):

L(+)-Ascorbic acid sodium salt, Vitamin C sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7NaO6
CAS Number:
Molecular Weight:
198.11
Beilstein:
3767246
EC Number:
MDL number:
UNSPSC Code:
12352205
eCl@ss:
34058010
PubChem Substance ID:
NACRES:
NA.75

biological source

synthetic (organic)

product line

BioReagent

Assay

≥98%

form

powder

mol wt

Mw 135.13 g/mol

technique(s)

cell culture | mammalian: suitable
cell culture | plant: suitable

color

white to off-white

mp

220 °C (dec.) (lit.)

solubility

H2O: 50 mg/mL

SMILES string

[Na+].OC[C@H](O)[C@H]1OC(=O)C(O)=C1[O-]

InChI

1S/C6H8O6.Na/c7-1-2(8)5-3(9)4(10)6(11)12-5;/h2,5,7-10H,1H2;/q;+1/p-1/t2-,5+;/m0./s1

InChI key

PPASLZSBLFJQEF-RXSVEWSESA-M

Looking for similar products? Visit Product Comparison Guide

General description

(+)-Sodium L-ascorbate (SA) is a potential antioxidant and is a derivative of vitamin C (L-ascorbic acid). It is widely used as a food additive. In addition, SA enhances growth retardation activity of bleomycin sulfate, R020-1724 (an inhibitor of cyclic nucleotide phosphodiesterase), prostaglandin E1 and sodium butyrate agents on neuroblastoma cells in culture. Elevated levels of SA induces two-stage urinary bladder carcinogenesis in rats. sodium L-ascorbate in combination with collagenogenesis inhibitors is used to treat dermal scars and keloids.

Application

(+)-Sodium L-ascorbate has been used:
  • to produce H2O2 in cell culture media
  • as a reducing agent for a cell-permeable artificial electron donor, N,N,N′,N′-tetramethyl-p-phenylenediamine (TMPD)
  • to study its effect on production of collagen and elastic fibers in fibroblasts cultures derived from normal human skin and dermal fat

Used for preparation of plant extracts; reductant for the isolation of chloroplasts.

Biochem/physiol Actions

Antioxidant

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Artefacts in cell culture: alpha-Ketoglutarate can scavenge hydrogen peroxide generated by ascorbate and epigallocatechin gallate in cell culture media
Long LH and Halliwell B
Biochemical and Biophysical Research Communications, 406(1), 20-24 (2011)
Artefacts in cell culture: pyruvate as a scavenger of hydrogen peroxide generated by ascorbate or epigallocatechin gallate in cell culture media
Long LH and Halliwell B
Biochemical and Biophysical Research Communications, 388(4), 700-704 (2009)
Promoting effects of sodium L-ascorbate on two-stage urinary bladder carcinogenesis in rats
Fukushima S, et al.
Cancer Research, 43(9), 4454-4457 (1983)
Sodium ascorbate potentiates the growth inhibitory effect of certain agents on neuroblastoma cells in culture
Prasad KN, et al.
Proceedings of the National Academy of Sciences of the USA, 76(2), 829-832 (1979)
The putative Drp1 inhibitor mdivi-1 is a reversible mitochondrial complex I inhibitor that modulates reactive oxygen species
Bordt EA, et al.
Developmental Cell, 40(6), 583-594 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service