Skip to Content
Merck
All Photos(1)

Key Documents

49752

Sigma-Aldrich

2-Phospho-L-ascorbic acid trisodium salt

≥95.0% (HPLC)

Synonym(s):

L-Ascorbic acid 2-phosphate trisodium salt, Sodium L-ascorbyl-2-phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6Na3O9P · xH2O
CAS Number:
Molecular Weight:
322.05 (anhydrous basis)
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.79

biological source

synthetic

Assay

≥95.0% (HPLC)

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

≤7.5% water (Karl Fischer)

color

white to off-white

pH

9.0-9.5 (25 °C, 30 g/L in H2O)

mp

260 °C

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤10 mg/kg
As: ≤2 mg/kg
Ba: ≤5 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤10 mg/kg
Mg: ≤10 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

[Na+].[Na+].[Na+].[P](=O)([O-])([O-])OC1=C([C@H](OC1=O)[C@@H](O)CO)[O-]

InChI

1S/C6H9O9P.3Na/c7-1-2(8)4-3(9)5(6(10)14-4)15-16(11,12)13;;;/h2,4,7-9H,1H2,(H2,11,12,13);;;/q;3*+1/p-3/t2-,4+;;;/m0.../s1

InChI key

YRWWOAFMPXPHEJ-OFBPEYICSA-K

Application

2-Phospho-L-ascorbic acid trisodium salt has been used as medium supplement in Dulbecco′s Modified Eagle Medium (DMEM) for adipose-derived stem cells, human mesenchymal stem cells and osteoblast cells.

Biochem/physiol Actions

L-Ascorbic acid 2-phosphate (AA2P) is used in biocatalytic dephosphorylation for electric power generation and electrochemical detection assays. L-Ascorbic acid 2-phosphate may be used in cell differentiation and tissue engineering applications. Asc-2P is used in studies of gene repression, such as the repression of dihydrotestosterone-induced dickkopf-1 expression.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Yang et al.
Journal of dental research, 96(2), 186-191 (2016-12-09)
In vitro expanded cell populations can contribute to bioengineered tooth formation but only as cells that respond to tooth-inductive signals. Since the success of whole tooth bioengineering is predicated on the availability of large numbers of cells, in vitro cell
Pericytes of multiple organs do not behave as mesenchymal stem cells in vivo
Guimaraes-Camboa N, et al.
Cell Stem Cell, 20(3), 345-359 (2017)
Visnja Radulovic et al.
Cell reports, 27(10), 2826-2836 (2019-06-06)
The distinct lineage potential is a key feature of hematopoietic stem cell (HSC) heterogeneity, but a subset of HSCs specialized for a single lymphoid compartment has not been identified. Here we report that HSCs expressing junctional adhesion molecule 2 (Jam2)
Regenerative therapy and cancer: in vitro and in vivo studies of the interaction between adipose-derived stem cells and breast cancer cells from clinical isolates
Zimmerlin L, et al.
Tissue Engineering: Part A, 17(1-2), 93-106 (2010)
Xiaojun Ji et al.
Life sciences, 231, 116674-116674 (2019-07-26)
Hypertrophic scar formation is a fibroproliferative disorder caused by abnormal wound healing. At present, there are limited treatment strategies for hypertrophic scars. In this study, we identified an endogenous peptide, LYENRL, through peptidomics screening that is downregulated in scar skin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service