Skip to Content
Merck
All Photos(5)

Documents

A3940

Sigma-Aldrich

1-Aminobenzotriazole

greener alternative

≥98% (TLC), powder, cytochrome P450 inhibitor

Synonym(s):

1-Benzotriazolamine, ABT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N4
CAS Number:
Molecular Weight:
134.14
Beilstein:
607843
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

1-Aminobenzotriazole,

Assay

≥98% (TLC)

Quality Level

form

powder

greener alternative product score

old score: 93
new score: 46
Find out more about DOZN™ Scoring

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Inherently Safer Chemistry for Accident Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

81-84 °C (lit.)

solubility

ethanol: 49-51 mg/mL, clear, pale yellow to yellow

greener alternative category

SMILES string

Nn1nnc2ccccc12

InChI

1S/C6H6N4/c7-10-6-4-2-1-3-5(6)8-9-10/h1-4H,7H2

InChI key

JCXKHYLLVKZPKE-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives. This product belongs to category of Re-engineered products, showing key improvements in Green Chemistry Principles “Waste Prevention”, “Safer Solvents and Auxiliaries” and “Inherently Safer Chemistry for Accident Prevention”. Click here to view its DOZN scorecard.

Application

1-Aminobenzotriazole has been used as a cytochrome P450 inhibitor to study the effects of estrogen on H2O2-induced premature senescence of vascular smooth muscle cells from old rats It has also been used to study the estrogen receptor-mediated effects of 17β-estradiol.

Biochem/physiol Actions

1-Aminobenzotriazole (1-ABT) is used as an in vitro tool in drug discovery. It can differentiate the P450 from non-P450 enzyme systems. 1-ABT exhibits inhibitory action by covalent alteration of the heme prosthetic group in the P450 enzyme.
Suicide inhibitor of cytochrome P450 and chloroperoxidase.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of estrogen on stress-induced premature senescence of vascular smooth muscle cells: a novel mechanism for the ?time window theory? of menopausal hormone therapy
Zhu C, et al.
Atherosclerosis, 215(2), 294-300 (2011)
M Lizeth Orozco Morales et al.
iScience, 25(1), 103571-103571 (2022-01-06)
Mesothelioma is a cancer that typically originates in the pleura of the lungs. It rapidly invades the surrounding tissues, causing pain and shortness of breath. We compared cell lines injected either subcutaneously or intrapleurally and found that only the latter
Is 1-aminobenzotriazole an appropriate in vitro tool as a nonspecific cytochrome P450 inactivator?
Linder CD, et al.
Drug Metabolism and Disposition, 37(1), 10-13 (2009)
Ailette Prieto et al.
Bioresource technology, 102(23), 10987-10995 (2011-09-23)
More than 90% of the antibiotics ciprofloxacin (CIPRO) and norfloxacin (NOR) at 2 mg L(-1) were degraded by Trametes versicolor after 7 days of incubation in malt extract liquid medium. In in vitro assays with purified laccase (16.7 nkat mL(-1))
Alex M Eddie et al.
ImmunoHorizons, 6(7), 432-446 (2022-07-12)
The type 2 cytokines IL-4 and IL-13, which share use of an IL-4 receptor α-chain and its nuclear induction of the transcription factor STAT6, are crucial in elicitation and maintenance of allergic conditions including asthma. STAT6 binds poly(ADP-ribose) polymerase (PARP)14

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service