Skip to Content
Merck
All Photos(2)

Key Documents

240907

Sigma-Aldrich

Trimethyl phosphite

≥99%

Synonym(s):

Methyl phosphite, P(OMe)3, Trimethoxyphosphine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)3P
CAS Number:
Molecular Weight:
124.08
Beilstein:
956570
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.3 (vs air)

Quality Level

vapor pressure

17 mmHg ( 20 °C)

Assay

≥99%

form

liquid

reaction suitability

reagent type: ligand
reaction type: Heck Reaction

reagent type: ligand
reaction type: Suzuki-Miyaura Coupling

refractive index

n20/D 1.408 (lit.)

bp

111-112 °C (lit.)

mp

−78 °C (lit.)

density

1.052 g/mL at 25 °C (lit.)

SMILES string

COP(OC)OC

InChI

1S/C3H9O3P/c1-4-7(5-2)6-3/h1-3H3

InChI key

CYTQBVOFDCPGCX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trimethyl phosphite is an organophosphorus compound commonly used as a polymerization catalyst, coating additive, and as an intermediate in the synthesis of flame retardants. In addition, it is also used as an activating reagent in amide formation.

Application

Trimethyl phosphite can be used as a ligand in the synthesis of allyliridium phosphite complexes by direct reaction with allyliridium 1,5-cyclooctadiene complex.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

82.4 °F - closed cup

Flash Point(C)

28 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Allyliridium phosphite complexes. Synthesis and chemistry
Muetterties EL, et al.
Organometallics, 1(12), 1562-1567 (1982)
An efficient protocol for the amidation of carboxylic acids promoted by trimethyl phosphite and iodine
Luo QL, et al.
European Journal of Organic Chemistry (2011)
N Ramanathan et al.
The journal of physical chemistry. A, 116(49), 12014-12023 (2012-11-21)
Trimethyl phosphite (TMPhite) and hydrogen chloride (HCl), when separately codeposited in a N(2) matrix, yielded a hydrogen bonded adduct, which was evidenced by shifts in the vibrational frequencies of the TMPhite and HCl submolecules. The structure and energy of the
Shinichi Yamabe et al.
The journal of physical chemistry. A, 114(43), 11699-11707 (2010-10-15)
Density functional theory calculations of the title reaction, P(OCH₃)₃ + (H₂O)(n) in CH₃CN, were conducted, where n is the number of water molecules. Two routes, the routes suggested by (A) Aksnes and (B) Arbuzov, were traced with various n values.
Bangwei Ding et al.
Journal of the American Chemical Society, 125(11), 3248-3259 (2003-03-13)
Trimethyl phosphite, (MeO)(3)P, is introduced as an efficient and selective trap in oxiranylcarbinyl radical (2) systems, formed from haloepoxides 8-13 under thermal AIBN/n-Bu(3)SnH conditions at about 80 degrees C. Initially, the transformations of 8-13, in the absence of phosphite, to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service