Skip to Content
Merck
All Photos(1)

Key Documents

SML0486

Sigma-Aldrich

Elacridar

≥98% (HPLC), powder, ABC transporters MDR-1 (P-gp) inhibitor

Synonym(s):

GF120918, GG918, GW0918, N-[4-[2-(3,4-Dihydro-6,7-dimethoxy-2(1H)-isoquinolinyl)ethyl]phenyl]-9,10-dihydro-5-methoxy-9-oxo-4-acridinecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H33N3O5
CAS Number:
Molecular Weight:
563.64
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.77

Product Name

Elacridar, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL (clear solution, warmed)

shipped in

wet ice

storage temp.

−20°C

SMILES string

COc1cccc2C(=O)c3cccc(C(=O)Nc4ccc(CCN5CCc6cc(OC)c(OC)cc6C5)cc4)c3Nc12

InChI

1S/C34H33N3O5/c1-40-28-9-5-7-26-32(28)36-31-25(33(26)38)6-4-8-27(31)34(39)35-24-12-10-21(11-13-24)14-16-37-17-15-22-18-29(41-2)30(42-3)19-23(22)20-37/h4-13,18-19H,14-17,20H2,1-3H3,(H,35,39)(H,36,38)

InChI key

OSFCMRGOZNQUSW-UHFFFAOYSA-N

Application

Elacridar has been used:
  • as a chemical to decipher the link between autophagy and the mechanism of resistance to sunitinib
  • as an ATP-binding cassette sub-family B member 1 inhibitor to study its effects on the NSC23766 cytotoxicity
  • to treat rhodamine 123 (Rh123) labeled cells to set sorting gates in fluorescence-activated cell sorting (FACS)

Biochem/physiol Actions

GF120918 (Elacridar) is a very potent inhibitor of the ABC transporters MDR-1 (P-gp) and BCRP. GF120918 increases the bioavailability of cytotoxic anti-tumor drugs, and also leads to increased levels of anti-HIV drugs in the brain and CNS.
GF120918 (Elacridar) is an inhibitor of MDR-1 (P-gp).

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The xenobiotic transporter Mdr1 enforces T cell homeostasis in the presence of intestinal bile acids
Cao W, et al.
Immunity, 47(6), 1182-1196 (2017)
Nico Holmstock et al.
Molecular pharmaceutics, 10(3), 1056-1062 (2013-01-31)
Rodent models are less suitable for predicting drug-drug interactions at the level of the human intestinal mucosa, especially when nuclear receptors such as pregnane X receptor (PXR) are involved. Recently, a transgenic mouse model, expressing both human PXR and CYP3A4
Hanneke G M Wittgen et al.
Molecular pharmaceutics, 9(5), 1351-1360 (2012-03-21)
Although the CB1 receptor antagonist/inverse agonist rimonabant has positive effects on weight loss and cardiometabolic risk factors, neuropsychiatric side effects have prompted researchers to develop peripherally acting derivatives. Here, we investigated for a series of 3,4-diarylpyrazoline CB1 receptor antagonists if
Resistance to sunitinib in renal clear cell carcinoma results from sequestration in lysosomes and inhibition of the autophagic flux
Giuliano S, et al.
Autophagy, 11(10), 1891-1904 (2015)
Seng Chuan Tang et al.
The Journal of pharmacology and experimental therapeutics, 341(1), 164-173 (2012-01-13)
N-desethyl sunitinib is a major and pharmacologically active metabolite of the tyrosine kinase inhibitor and anticancer drug sunitinib. Because the combination of N-desethyl sunitinib and sunitinib represents total active drug exposure, we investigated the impact of several multidrug efflux transporters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service