Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ammonium nitrate
Synonym(s):
Ammonium nitrate
Linear Formula:
NH
4
NO
3
CAS Number:
6484-52-2
Molecular Weight:
80.04
EC Number:
229-347-8
Products
Product Category
salts (6)
Brand
Sigma-Aldrich (5)
Supelco (2)
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
A9642
Ammonium nitrate,
≥99.0%
Expand
View Pricing
221244
Ammonium nitrate,
ACS reagent, ≥98%
Expand
View Pricing
1.01188
Ammonium nitrate,
for analysis EMSURE
®
ACS
Expand
View Pricing
A3795
Ammonium nitrate,
suitable for plant cell culture
Expand
View Pricing
1.01187
Ammonium nitrate,
EMPLURA
®
Expand
View Pricing
256064
Ammonium nitrate,
99.999% trace metals basis
Expand
View Pricing
A7455
Ammonium nitrate,
BioXtra, ≥99.5%
Expand
View Pricing