Skip to Content
Merck
All Photos(2)

Key Documents

A2306

Sigma-Aldrich

Monoclonal Anti-Rabbit Immunoglobulins−Alkaline Phosphatase antibody produced in mouse

clone RG-16, purified immunoglobulin, buffered aqueous glycerol solution

Synonym(s):

Monoclonal Anti-Rabbit Immunoglobulins (IgG, IgA, IgM)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

conjugate

alkaline phosphatase conjugate

antibody form

purified immunoglobulin

antibody product type

secondary antibodies

clone

RG-16, monoclonal

form

buffered aqueous glycerol solution

species reactivity

rabbit

should not react with

pig, chicken, bovine, horse, human, guinea pig, turkey, rat, canine, goat, feline, sheep

technique(s)

direct ELISA: 1:40,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:160 using human tonsil and Cat. No. I8635 as the primary antibody
western blot: 1:160,000-1:320,000 using detecting β-actin in total cell extract of HeLa cells (5-10 μg per well)

isotype

IgG1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Monoclonal Anti-Rabbit Immunoglobulins (mouse IgG1 isotype) is derived from the hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse. Immunoglobulins is composed of two heavy (H) and two light (L) chains. Each containing one NH2-terminal variable region and one or more COOH-terminal constant regions. They are heterodimeric proteins.
Reacts with an epitope on the heavy chain of rabbit IgG, IgA and IgM, which is sensitive to reduction. No cross-reactivity with human IgG, IgA and IgM.

Specificity

Binds only rabbit Igs. Does not react with reduced rabbit Igs.
No cross reaction is observed with IgG from the following species: bovine, cat, chicken, dog, goat, guinea pig, horse, pig, rat, sheep or turkey.

Application

Monoclonal Anti-Rabbit Immunoglobulins Alkaline Phosphatase antibody produced in mouse has been used in:
  • indirect enzyme linked immunosorbent assay (ELISA)
  • immunoblotting
  • immunohistochemistry

The mouse monoclonal anti-Rabbit IgG (heavy chain-specific) peroxidase conjugated antibody can be used as a secondary antibody for western blot analysis of protein preps from E. coli cultures at a concentration of 1:10000 diluted in PBSt for 1 hour at room temperature.

Biochem/physiol Actions

IgGs are glycoprotein antibodies that modulate several immune responses. Rabbit IgGs against target proteins are often used as primary antibodies in various research applications. Thus, secondary anti-rabbit IgGs conjugated to a detectable substrate are useful tools for the analysis of target proteins.
Mouse monoclonal anti-rabbit immunoglobulins-alkaline phosphatase antibody binds to an epitope on the heavy chain of rabbit IgG, IgA, and IgM. The product stains bands at the intact whole molecule and at the heavy chains in an immunoblot of denatured, non-reduced rabbit immunoglobulins. Reduction of rabbit immunoglobulins destroys the epitope. In ELISA, the product shows no cross reaction with human serum or tissue preparations, nor with human IgA, IgG, or IgM.
The constant domains of immunoglobulins are involved in the complement activation.

Physical form

Solution in 0.05 M Tris buffer pH 8.0, containing 1 mM MgCl2, 0.1% bovine serum albumin, 50% glycerol and 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The immune evasion paradox: immunoevasins of murine cytomegalovirus enhance priming of CD8 T cells by preventing negative feedback regulation
Bohm V.et al.
Journal of virology, 82(23), 11637-11650 (2008)
Yasumasa Okazaki et al.
Cancer science, 104(8), 989-995 (2013-04-30)
Malignant mesothelioma (MM), which is associated with asbestos exposure, is one of the most deadly tumors in humans. Early MM is concealed in the serosal cavities and lacks specific clinical symptoms. For better treatment, early detection and prognostic markers are
Polymerization of nonfilamentous actin into microfilaments is an important process for porcine oocyte maturation and early embryo development
Wang WH, et al.
Biology of Reproduction, 62(5), 1177-1183 (2000)
Till Myrach et al.
The Journal of biological chemistry, 292(35), 14556-14565 (2017-07-16)
Urease is a ubiquitous nickel metalloenzyme. In plants, its activation requires three urease accessory proteins (UAPs), UreD, UreF, and UreG. In bacteria, the UAPs interact with urease and facilitate activation, which involves the channeling of two nickel ions into the
Ewa Wiśniewska-Chudy et al.
Oncology reports, 37(4), 2295-2307 (2017-03-06)
Myeloproliferative neoplasms (MPNs) are clonal hematopoietic progenitor cell disorders characterized by the proliferation of one or more hematopoietic lineages. The classical MPNs include polycythemia vera (PV), essential thrombocythemia (ET) and primary myelofibrosis (PMF) entities. These disorders are characterized by bone

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service