Skip to Content
Merck
All Photos(1)

Documents

AP132A

Sigma-Aldrich

Goat Anti-Rabbit IgG Antibody, Alkaline Phosphatase conjugate

0.6 mg/mL, Chemicon®

Synonym(s):

Alkaline Phosphatase Anti-Rabbit, Anti-Rabbit IgG AP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

alkaline phosphatase conjugate

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

rabbit

manufacturer/tradename

Chemicon®

concentration

0.6 mg/mL

technique(s)

ELISA: suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

General description

Goat anti Rabbit IgG affinity purified antibody, Alkaline Phosphatase conjugated.
Immunoglobulin G (IgG), is one of the most abundant proteins in human serum with normal levels between 8-17 mg/mL in adult blood. IgG is important for our defence against microorganisms and the molecules are produced by B lymphocytes as a part of our adaptive immune response. The IgG molecule has two separate functions; to bind to the pathogen that elicited the response and to recruit other cells and molecules to destroy the antigen. The variability of the IgG pool is generated by somatic recombination and the number of specificities in an individual at a given time point is estimated to be 1011 variants.

Application

ELISA: 1:5,000-1:50,000

Western Blot: 1:5,000-1:50,000

Optimal working dilutions must be determined by the end user.
Goat anti-Rabbit IgG Antibody, Alkaline Phosphatase conjugate is an antibody against Rabbit IgG for use in ELISA & WB.
Research Category
Secondary & Control Antibodies
Research Sub Category
Whole Immunoglobulin Secondary Antibodies

Linkage

Replaces: AP158A

Physical form

ImmunoAffinity Purified
RECONSTITUTION:

Reconstitute with sterile distilled water to match the volume indicated on the vial label. Centrifuge or vortex this product if it is not completely clear after standing for 1-2 hours at room temperature.

Storage and Stability

Store lyophilized material at 2-8°C until opened. After reconstitution this product is stable for several weeks at 2-8°C as an undiluted liquid. DO NOT FREEZE. For extended storage after reconstitution, add an equal volume of glycerol to make a final concentration of 50% glycerol and store at 2-8°C for up to 12 months. Please note that the concentration of protein and buffer salts will decrease to one-half of the original after adding glycerol.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chia-Jui Liu et al.
PloS one, 14(2), e0212909-e0212909 (2019-03-01)
In Klebsiella pneumoniae CG43S3, deletion of the response regulator gene rcsB reduced the capsular polysaccharide amount and survival on exposure to acid stress. A comparison of the pH 4.4-induced proteomes between CG43S3 and CG43S3ΔrcsB revealed numerous differentially expressed proteins and
Joo-Yeun Oh et al.
Transfusion, 55(12), 2967-2978 (2015-07-24)
Storage-dependent damage to red blood cells (RBCs) varies significantly. Identifying RBC units that will undergo higher levels of hemolysis during storage may allow for more efficient inventory management decision-making. Oxidative-stress mediates storage-dependent damage to RBCs and will depend on the
Jon A Detterich et al.
Free radical biology & medicine, 141, 408-415 (2019-07-07)
Sickle cell disease (SCD) is a monogenetic disease that results in the formation of hemoglobin S. Due to more rapid oxidation of hemoglobin S due to intracellular heme and adventitious iron in SCD, it has been thought that an inherent
Erin E Burns et al.
Pesticide biochemistry and physiology, 140, 69-78 (2017-08-02)
Extensive herbicide usage has led to the evolution of resistant weed populations that cause substantial crop yield losses and increase production costs. The multiple herbicide resistant (MHR) Avena fatua L. populations utilized in this study are resistant to members of
Victoria M Harper et al.
Antioxidants & redox signaling, 22(4), 294-307 (2014-09-30)
Transfusion with stored red blood cells (RBCs) is associated with increased morbidity and mortality. Peroxiredoxin-2 (Prx-2) is a primary RBC antioxidant that limits hydrogen peroxide (H2O2)-mediated toxicity. Whether Prx-2 activity is altered during RBC storage is not known. Basal and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service