Skip to Content
Merck

Skip To

A3673

Anti-Mouse IgG (γ-chain specific)−Peroxidase antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Goat Anti-Mouse IgG (γ-chain specific)−HRP

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
MDL number:
Conjugate:
peroxidase conjugate
Clone:
polyclonal
Application:
ELISA (d)
Citations:
85
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

goat

Quality Segment

conjugate

peroxidase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse

technique(s)

direct ELISA: 1:15,000

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Immunoglobulin G (IgG) is a glycoprotein antibody that regulates immune responses such as phagocytosis and is also involved in the development of autoimmune diseases. Mouse IgGs have four distinct isotypes, namely, IgG1, IgG2a, IgG2b, and IgG3. IgG1 regulates complement fixation in mice.
Goat Anti-Mouse IgG (γ-chain specific)-Peroxidase antibody is specific for mouse IgG when tested against purified mouse IgA, IgG, and IgM myeloma proteins.

Immunogen

purified mouse IgG

Application

Goat Anti-Mouse IgG (γ-chain specific)-Peroxidase antibody has been used for ELISA applications at a dilution of 1:3000.
HRP conjugated goat anti-mouse gamma chain specific was used to detect neutralizing antibodies against HIV1 in sera and mucosal washings by ELISA. 3′3′5,5 tetramethylbenzidine (Sigma) was used as the substrate for development.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin with preservative.

Preparation Note

Prepared using the periodate method described by Wilson, M.B., and Nakane, P.K., in Immunofluorescence and Related Staining Techniques, Elsevier/North Holland Biomedical Press, Amsterdam, p215 (1978).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A6029A0420A3187
biological source

goat

biological source

goat

biological source

goat

biological source

goat

species reactivity

mouse

species reactivity

human

species reactivity

human

species reactivity

human

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

technique(s)

direct ELISA: 1:15,000

technique(s)

direct ELISA: 1:10,000

technique(s)

direct ELISA: 1:50,000, dot blot: 1:100,000 (chemiluminescent), immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:200

technique(s)

direct ELISA: 1:30,000, immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50, western blot: 1:30,000

conjugate

peroxidase conjugate

conjugate

peroxidase conjugate

conjugate

peroxidase conjugate

conjugate

alkaline phosphatase conjugate

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal


Still not finding the right product?

Try our Product Selector Tool to narrow your options


pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content






Global Trade Item Number

SKUGTIN
A3673-.25ML04061835805037
A3673-.5ML04061835805044
A3673-1ML04061833367773

Questions

Reviews

No rating value

Active Filters