Skip to Content
Merck
All Photos(1)

Key Documents

A3626

Sigma-Aldrich

Nα-Acetyl-L-ornithine

≥98% (TLC), suitable for ligand binding assays

Synonym(s):

N2-acetyl-L-lysine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14N2O3
CAS Number:
Molecular Weight:
174.20
MDL number:
UNSPSC Code:
12352202
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

Product Name

Nα-Acetyl-L-ornithine,

Assay

≥98% (TLC)

form

powder

technique(s)

ligand binding assay: suitable

color

colorless to white

storage temp.

−20°C

SMILES string

CC(=O)N[C@@H](CCCN)C(O)=O

InChI

1S/C7H14N2O3/c1-5(10)9-6(7(11)12)3-2-4-8/h6H,2-4,8H2,1H3,(H,9,10)(H,11,12)/t6-/m0/s1

InChI key

JRLGPAXAGHMNOL-LURJTMIESA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Nα-Acetyl-L-ornithine (AORN) is a substrate for the identification, differentiation and characterization of N(α)-acetyl-L-ornithine deacetylase(s) and of N-Acetyl-l-ornithine transcarbamylase(s) (AOTCase) found in plants, some eubacteria and some human pathogens.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nadia Botini et al.
Biochimica et biophysica acta. Proteins and proteomics, 1869(2), 140561-140561 (2020-11-09)
Somatic embryogenesis is an important biotechnological technique for large-scale propagation of elite genotypes. Identifying stage-specific compounds associated with somatic embryo development can help elucidate the ontogenesis of Carica papaya L. somatic embryos and improve tissue culture protocols. To identify the
Séverine Lemarié et al.
Plant & cell physiology, 56(11), 2158-2168 (2015-09-13)
The role of salicylic acid (SA) and jasmonic acid (JA) signaling in resistance to root pathogens has been poorly documented. We assessed the contribution of SA and JA to basal and partial resistance of Arabidopsis to the biotrophic clubroot agent
Yongdong Li et al.
Biochemistry, 49(32), 6887-6895 (2010-08-11)
N-Acetyl-l-ornithine transcarbamylase (AOTCase), rather than ornithine transcarbamylase (OTCase), is the essential carbamylase enzyme in the arginine biosynthesis of several plant and human pathogens. The specificity of this unique enzyme provides a potential target for controlling the spread of these pathogens.
Dashuang Shi et al.
Proteins, 64(2), 532-542 (2006-06-03)
N-acetyl-L-ornithine transcarbamoylase (AOTCase) is a new member of the transcarbamoylase superfamily that is essential for arginine biosynthesis in several eubacteria. We report here crystal structures of the binary complexes of AOTCase with its substrates, carbamoyl phosphate (CP) or N-acetyl-L-ornithine (AORN)
Daniel Charlier et al.
Amino acids, 51(8), 1103-1127 (2019-07-04)
Already very early, the study of microbial arginine biosynthesis and its regulation contributed significantly to the development of new ideas and concepts. Hence, the term "repression" was proposed by Vogel (The chemical basis of heredity, The John Hopkins Press, Baltimore

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service