Skip to Content
Merck
All Photos(1)

Documents

114286

Sigma-Aldrich

4-Hydroxyphenylpyruvic acid

98%

Synonym(s):

3-(4-Hydroxyphenyl)-2-oxopropanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CH2COCO2H
CAS Number:
Molecular Weight:
180.16
Beilstein:
2691632
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

219-220 °C (dec.) (lit.)

solubility

ethanol: soluble 50 mg/mL

SMILES string

OC(=O)C(=O)Cc1ccc(O)cc1

InChI

1S/C9H8O4/c10-7-3-1-6(2-4-7)5-8(11)9(12)13/h1-4,10H,5H2,(H,12,13)

InChI key

KKADPXVIOXHVKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-hydroxyphenylpyruvic acid can be determined in pork meat and Iberian ham samples by a sensitive method of multiple reaction monitoring (MRM) by mass spectrometry.

Application

<ul>
<li><strong>Identification of serum biomarkers of ischemic stroke:</strong>4-Hydroxyphenylpyruvic acid is used as a potential diagnostic biomarker in the study to distinguish hypertensive ischemic stroke (IS) patients from both healthy individuals and those with hypertension (Zhao et al., 2023).</li>
</ul>

Preparation Note

50 gm of 4-Hydroxyphenylpyruvic acid dissolves in 1 mL of ethanol to yield a clear, light yellow solution.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Francisco J Hidalgo et al.
Food chemistry, 140(1-2), 183-188 (2013-04-13)
An analytical method which offers accurate determination and identification of eight α-keto acids (α-ketoglutaric acid, pyruvic acid, 4-hydroxyphenylpyruvic acid, 3-methyl-2-oxobutyric acid, α-keto-γ-methylthiobutyric acid, 4-methyl-2-oxovaleric acid, 3-methyl-2-oxovaleric acid, and phenylpyruvic acid) in pork meat and Iberian ham samples is reported. The
Etresia van Dyk et al.
Biochemical and biophysical research communications, 338(2), 815-819 (2005-11-03)
Tyrosinemia type 1 (HT1) is an autosomal recessive disorder of the tyrosine metabolism in which the fumarylacetoacetate hydrolase enzyme is defective. This disease is clinically heterogeneous and a chronic and acute form is discerned. Characteristic of the chronic form is
Márcia Eliana da Silva Ferreira et al.
Eukaryotic cell, 5(8), 1441-1445 (2006-08-10)
Mutations in the human HPD gene (encoding 4-hydroxyphenylpyruvic acid dioxygenase) cause hereditary tyrosinemia type 3 (HT3). We deleted the Aspergillus nidulans homologue (hpdA). We showed that the mutant strain is not able to grow in the presence of phenylalanine and
Yuwu Chi et al.
Bioelectrochemistry (Amsterdam, Netherlands), 60(1-2), 37-45 (2003-08-02)
The keto-enol tautomerization of p-hydroxyphenylpyruvic acid (pHPP) in aqueous solutions and the complexation reaction between enolic pHPP and boric acid have been studied by electrochemical techniques including linear sweep voltammetry (LSV), pulse voltammetry, and cyclic voltammetry (CV), combining with UV
Tomasz Borowski et al.
Biochemistry, 43(38), 12331-12342 (2004-09-24)
Density functional calculations using the B3LYP functional has been used to study the reaction mechanism of 4-hydroxyphenylpyruvate dioxygenase. The first part of the catalytic reaction, dioxygen activation, is found to have the same mechanism as in alpha-ketoglutarate-dependent enzymes; the ternary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service