Skip to Content
Merck
All Photos(1)

Documents

A3525

Sigma-Aldrich

Anti-Human IgE (ε-chain specific)−Alkaline Phosphatase antibody produced in goat

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Goat Anti-Human IgE (ε-chain specific)−AP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

alkaline phosphatase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

direct ELISA: 1:7,000-1:21,000

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Affinity isolated antibody is obtained from anti-human IgE antiserum by immunospecific purification which removes essentially all goat serum proteins, including immunoglobulins, which do not specifically bind to the ε-chain of human IgE. Anti-Human IgE is conjugated to alkaline phosphatase by protein cross linking with 0.2% glutaraldehyde. IgE is present in low levels compared to other immunoglobulin isotypes. It exists in two forms, one being a secreted by plasma cell and the other form is associated with the B cell receptor.
IgEs are glycoprotein antibodies that regulate immunological responses to allergies and infections. These immunoglobulins have been implicated in mediating Type I hypersensitivity reactions. Elevated IgE levels have been associated with hyper IgE syndrome and allergic asthma . Anti-Human IgE (epsilon-chain specific)-Alkaline Phosphatase antibody is specific for human IgE when tested against human IgA, IgG, IgM, Bence Jones κ and λ myeloma proteins.

Immunogen

Purified human IgE

Application

Anti-Human IgE (ε-chain specific)−Alkaline Phosphatase antibody produced in goat has been used in dot blot.
Anti-Human IgE (epsilon-chain specific)-Alkaline Phosphatase antibody is suitable for use in immunoblotting and ELISA (at 1:500 dilutions using 100μ/well).
IgE secreted from transfected murin myeloma cells was detected by ELISA using the alkaline phosphatase-conjugated goat anti-human IgE (epsilon specific) antibody.

Biochem/physiol Actions

IgE bound to B cell receptor mediates antigen uptake and regulates differentiation and proliferation of B cells.

Physical form

Solution in 0.05 M Tris, pH 8.0, containing 1% bovine serum albumin, 1 mM MgCl2, 15 mM sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dalgys Martínez et al.
Immunology letters, 196, 103-112 (2018-02-07)
Sensitization to allergens of the house dust mites Dermatophagoides pteronyssinnus and Blomia tropicalis is an important risk factor for asthma and allergic diseases. Allergen-specific immunotherapy is currently based on natural allergen extracts, however, in the last years recombinant allergens with
Jack H Jhamandas et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 24(24), 5579-5584 (2004-06-18)
Salvage of cholinergic neurons in the brain through a blockade of the neurotoxic effects of amyloidbeta protein (Abeta) is one of the major, but still elusive, therapeutic goals of current research in Alzheimer's disease (AD). To date, no receptor has
Serum proteins and fractions, HDL-cholesterol and total IgG and IgE levels in cases of acute and chronic paracoccidioidomycosis.
Marquez, Ade, S., et al.
Rev. Biochem., 42(3), 245-249 (2009)
C D Lindstrøm et al.
Scandinavian journal of immunology, 44(4), 335-344 (1996-10-01)
Allergy to fish is common in Northern Europe. Variable reactions to different fish species are usually experienced among fish allergic patients. The allergens of cod fish and particularly the major allergen parvalbumin beta (Gadus callarias) have been extensively studied in
James A Gregory et al.
Plant biotechnology journal, 14(7), 1541-1550 (2016-01-24)
Peanut allergy is an IgE-mediated adverse reaction to a subset of proteins found in peanuts. Immunotherapy aims to desensitize allergic patients through repeated and escalating exposures for several months to years using extracts or flours. The complex mix of proteins

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service