Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
biological source
rabbit
Quality Level
conjugate
unconjugated
antibody form
whole antiserum
antibody product type
primary antibodies
clone
polyclonal
mol wt
antigen 66.437-66.6 kDa
contains
15 mM sodium azide
species reactivity
human
technique(s)
indirect ELISA: 1:60,000, quantitative precipitin assay: 2.4-3.6
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... ALB(213)
General description
Immunogen
Application
Biochem/physiol Actions
Physical form
1 of 1
This Item | |||
|---|---|---|---|
| biological source rabbit | biological source chicken | biological source rabbit | biological source chicken |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| Quality Level 200 | Quality Level 100 | Quality Level 200 | Quality Level 200 |
| antibody form whole antiserum | antibody form affinity isolated antibody | antibody form whole antiserum | antibody form affinity isolated antibody |
| species reactivity human | species reactivity human, rat, mouse | species reactivity chicken | species reactivity human, rat, mouse |
| Gene Information human ... ALB(213) | Gene Information human ... ALB(213) | Gene Information chicken ... Alb(396197) | Gene Information human ... ALB(213) |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
13 - Non Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Instructions
Global Trade Item Number
| SKU | GTIN |
|---|---|
| A3293-1ML | 04061837856129 |



