Skip to Content
Merck
All Photos(1)

Key Documents

GW10067F

Sigma-Aldrich

Anti-Albumin (serum) antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Serum albumin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, rat, mouse

manufacturer/tradename

Genway 15-288-10067F

technique(s)

indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ALB(213)

General description

Serum albumin is the major protein component of blood produced in liver. It regulates colloid osmotic blood pressure, transportation of a variety of substances, blood pH and facilitates an endogenous source of amino acids. It also facilitates the transportation of zinc in plasma. Anti-albumin (serum) antibody (diluted 1: 200) can be used in staining cells or tissues. Chicken anti-albumin (serum) antibody reacts specifically with serum albumin.

Immunogen

Immunogen Sequence: GI # 113576, sequence 1-609
Recombinant protein Serum albumin.

Application

Anti-albumin (serum) antibody can be used in indirect ELISA and western blotting (diluted 1: 500).

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jin Lu et al.
Biochemical Society transactions, 36(Pt 6), 1317-1321 (2008-11-22)
Although details of the molecular mechanisms for the uptake of the essential nutrient zinc into the bloodstream and its subsequent delivery to zinc-requiring organs and cells are poorly understood, it is clear that in vertebrates the majority of plasma zinc

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service