Skip to Content
Merck
All Photos(2)

Key Documents

A1949

Sigma-Aldrich

Anti-Rabbit IgG (γ-chain specific)–Peroxidase antibody, Mouse monoclonal

clone RG-96, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-Rabbit IgG (γ-chain specific)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

Quality Level

conjugate

peroxidase conjugate

antibody form

purified immunoglobulin

antibody product type

secondary antibodies

clone

RG-96, monoclonal

form

lyophilized powder

species reactivity

rabbit

should not react with

goat, feline, pig, guinea pig, rat, bovine, canine, human, horse, sheep, chicken

packaging

vial of 0.5 mL

technique(s)

direct ELISA: 1:40,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:200
western blot: 1:160,000 using total cell extract of HeLa cells (5-10 μg/well)

isotype

IgG1

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Anti-Rabbit IgG (γ-chain specific)-Peroxidase antibody, Mouse monoclonal is a monoclonal anti-rabbit IgG conjugated to horseradish peroxidase. Monoclonal antirabbit IgG is derived from the RG-96 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from mice immunized with rabbit IgG.
Rabbit IgGs are often used as primary antibodies in several immunological assays. Thus, polyclonal anti-rabbit IgGs are usually used as secondary antibodies in such applications. However, polyclonal antibodies to rabbit IgGs can also cross-react with IgGs from other species, especially with human IgGs, and thereby lead to a lack of specificity in test results. The use of monoclonal anti-rabbit IgG that is specific for only rabbit IgG improves the accuracy of immunological assays and thus, can be useful for various research applications.
The product recognizes an epitope located on the γ (heavy)-chain of rabbit IgG. In immunoblotting, the antibody recognizes both native and denatured forms of rabbit IgG. In ELISA, the antibody is specific for rabbit IgG, and shows no cross-reactivity with rabbit IgA and IgM or human IgG, IgA, and IgM. No cross reaction is observed with IgG from the following species: bovine, cat, chicken, dog, goat, guinea pig, horse, pig, rat, or sheep.

Immunogen

Rabbit IgG

Application

Monoclonal Anti-Rabbit IgG (γ-chain specific)- Peroxidase has been used in
  • various immunochemical assays like enzyme linked immunosorbent assay (ELISA)
  • westernblotting
  • immunohistology
  • dot immunobinding assay

Physical form

Lyophilized from 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 0.05% MIT.

Preparation Note

Prepared by the two-step glutaraldehyde method described by Avrameas, S., et al., Scand. J. Immunol., 8, Suppl. 7, 7 (1978).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

NsrR: a key regulator circumventing Salmonella enterica serovar Typhimurium oxidative and nitrosative stress in vitro and in IFN-gamma-stimulated J774. 2 macrophages
Gilberthorpe NJ, et al.
Microbiology, 153(Pt 6), 1756-1756 (2007)
Michal T Boniecki et al.
Nature communications, 8(1), 1287-1287 (2017-11-04)
Iron-sulfur (Fe/S) clusters are essential protein cofactors crucial for many cellular functions including DNA maintenance, protein translation, and energy conversion. De novo Fe/S cluster synthesis occurs on the mitochondrial scaffold protein ISCU and requires cysteine desulfurase NFS1, ferredoxin, frataxin, and
Yundan Wang et al.
PLoS pathogens, 9(1), e1003102-e1003102 (2013-01-18)
The stress of living conditions, similar to infections, alters animal immunity. High population density is empirically considered to induce prophylactic immunity to reduce the infection risk, which was challenged by a model of low connectivity between infectious and susceptible individuals
Leo Heyndrickx et al.
PloS one, 8(9), e74552-e74552 (2013-09-12)
Ten to 30% of HIV-1 infected subjects develop broadly neutralizing antibodies (bNAbs) during chronic infection. We hypothesized that immunizing rabbits with viral envelope glycoproteins (Envs) from these patients may induce bNAbs, when formulated as a trimeric protein and in the
Targeted inhibition of EG-1 blocks breast tumor growth
Lu M, et al.
Cancer Biology & Therapy, 6(6), 936-941 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service