Skip to Content
Merck
All Photos(3)

Key Documents

A1784

Sigma-Aldrich

Aminopterin

powder

Synonym(s):

(S)-2-{4-[(2,4-Diaminopteridin-6-yl)methylamino]benzamido}pentanedioic acid, 4-Amino-PGA, 4-Aminofolic acid, 4-Aminopteroyl-L-glutamic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20N8O5
CAS Number:
Molecular Weight:
440.41
Beilstein:
69045
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

Assay

≥97% (TLC)

form

powder

color

yellow

mp

225 °C

solubility

2 M NaOH: 50 mg/mL (Solutions may be stored for 1-2 days at 4°C.)
DMSO: soluble (Solutions may be stored for 1-2 days at 4°C.)

ε (extinction coefficient)

24,500 at 282 nm in 0.1 M NaOH at 1 M
25,700 at 261 nm in 0.1 M NaOH at 1 M
8,100 at 373 nm in 0.1 M NaOH at 1 M

storage temp.

−20°C

SMILES string

Nc1nc(N)c2nc(CNc3ccc(cc3)C(=O)N[C@@H](CCC(O)=O)C(O)=O)cnc2n1

InChI

1S/C19H20N8O5/c20-15-14-16(27-19(21)26-15)23-8-11(24-14)7-22-10-3-1-9(2-4-10)17(30)25-12(18(31)32)5-6-13(28)29/h1-4,8,12,22H,5-7H2,(H,25,30)(H,28,29)(H,31,32)(H4,20,21,23,26,27)/t12-/m0/s1

InChI key

TVZGACDUOSZQKY-LBPRGKRZSA-N

Gene Information

human ... FPGS(2356)
mouse ... Fpgs(14287)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Folic acid antagonist that binds to dihydrofolate reductase and inhibits its activity.
Folic acid antagonist. Aminopterin is actively transported into cells by the folate transporter. In the cell, it is converted to a high molecular weight polyglutamate metabolite by folylpolyglutamate synthase that, in turn, binds to dihydrofolate reductase and inhibits its activity. Aminopterin-polyglutamate is degraded intracellularly by γ-glutamyl hydrolase.

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Linkage

More potent, but more toxic, than methotrexate.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Oral - Eye Irrit. 2 - Muta. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Min Zhang et al.
Cell, 181(3), 637-652 (2020-04-10)
Many cytosolic proteins lacking a signal peptide, called leaderless cargoes, are secreted through unconventional secretion. Vesicle trafficking is a major pathway involved. It is unclear how leaderless cargoes enter into the vesicle. Here, we find a translocation pathway regulating vesicle
Aleem Gangjee et al.
Journal of medicinal chemistry, 48(16), 5329-5336 (2005-08-05)
We report, for the first time, the biological activities of four-carbon-atom bridged classical antifolates on dihydrofolate reductase (DHFR), thymidylate synthase (TS), and folylpolyglutamate synthetase (FPGS) as well as antitumor activity. Extension of the bridge homologation studies of classical two-carbon bridged
A Rosowsky
Current medicinal chemistry, 6(4), 329-352 (1999-04-02)
Nonpolyglutamatable antifolates are potentially of therapeutic interest for the treatment of tumors that are inherently refractory, or have become resistant, to classical antifolates as a result of decreased expression of the enzyme folylpolyglutamate synthetase. An interesting class of water-soluble nonpolyglutamatable
M Del Campo et al.
Teratology, 60(1), 10-12 (1999-07-21)
Maternal exposures to aminopterin and methotrexate have been associated with a pattern of malformation which includes prenatal-onset growth deficiency, severe lack of ossification of the calvarium, hypoplastic supraorbital ridges, small, low-set ears, micrognathia, and limb abnormalities. We report on a
Hatem A Azim et al.
Current opinion in oncology, 24(3), 325-331 (2012-02-22)
To discuss the clinical development of new formulations of old cytotoxic agents and highlight the value of adopting this strategy. Several drugs are currently in clinical development with high potential in improving clinical outcomes compared with their older counterparts. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service