Skip to Content
Merck
All Photos(1)

Documents

M1000000

Methotrexate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(+)-Amethopterin, 4-Amino-10-methylfolic acid, 4-Amino-N10-methylpteroyl-L-glutamic acid, Antifolan, MTX, Methylaminopterin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22N8O5
CAS Number:
Molecular Weight:
454.44
Beilstein:
70669
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

methotrexate

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CN(Cc1cnc2nc(N)nc(N)c2n1)c3ccc(cc3)C(=O)N[C@@H](CCC(O)=O)C(O)=O

InChI

1S/C20H22N8O5/c1-28(9-11-8-23-17-15(24-11)16(21)26-20(22)27-17)12-4-2-10(3-5-12)18(31)25-13(19(32)33)6-7-14(29)30/h2-5,8,13H,6-7,9H2,1H3,(H,25,31)(H,29,30)(H,32,33)(H4,21,22,23,26,27)/t13-/m0/s1

InChI key

FBOZXECLQNJBKD-ZDUSSCGKSA-N

Gene Information

human ... DHFR(1719)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Methotrexate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Muta. 2 - Repr. 1B - STOT RE 1

Target Organs

Liver,Bone marrow

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Driehuis et al.
PloS one, 15(5), e0231588-e0231588 (2020-05-19)
We have recently established a protocol to grow wildtype human oral mucosa organoids. These three-dimensional structures can be maintained in culture long-term, do not require immortalization, and recapitulate the multilayered composition of the epithelial lining of the oral mucosa. Here
Axel Finckh et al.
Arthritis research & therapy, 16(5), 458-458 (2014-10-16)
Calcium pyrophosphate deposition (CPPD) may cause severe arthropathy, major joint destruction and treatment options are limited. The aim of this study was to test the therapeutic efficacy of methotrexate (MTX) in chronic or recurrent CPPD arthropathy. Patients with CPPD arthropathy
Anna-Birgitte Aga et al.
Annals of the rheumatic diseases, 74(2), 381-388 (2013-11-29)
To investigate whether baseline disease activity levels and responses in patients with rheumatoid arthritis (RA) changed during the period 2000-2010. Data were provided by the Norwegian disease-modifying antirheumatic drug (NOR-DMARD) study. Patients with inflammatory joint diseases starting new treatment with
Joshua F Baker et al.
Annals of the rheumatic diseases, 73(11), 1968-1974 (2013-08-02)
To determine if early MRI measures predict X-ray progression at 1 and 2 years in a large RA trial cohort. This study included 256 methotrexate (MTX)-naïve RA patients from a randomised placebo-controlled trial of golimumab (GO-BEFORE). MRIs of wrist and 2nd-5th
Giovanni L Mancardi et al.
Neurology, 84(10), 981-988 (2015-02-13)
To assess in multiple sclerosis (MS) the effect of intense immunosuppression followed by autologous hematopoietic stem cells transplantation (AHSCT) vs mitoxantrone (MTX) on disease activity measured by MRI. We conducted a multicenter, phase II, randomized trial including patients with secondary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service