Skip to Content
Merck
All Photos(2)

Documents

D7006

Sigma-Aldrich

Dihydrofolic acid

≥90%

Synonym(s):

7,8-Dihydropteroyl-L-glutamic acid, Dihydropteroyl-L-glutamic acid, FAH2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21N7O6
CAS Number:
Molecular Weight:
443.41
Beilstein:
69017
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.79

biological source

synthetic (organic)

Quality Level

Assay

≥90%

form

powder

color

light yellow to dark yellow-orange

storage temp.

−20°C

SMILES string

NC1=NC(=O)C2=C(NCC(CNc3ccc(cc3)C(=O)N[C@@H](CCC(O)=O)C(O)=O)=N2)N1

InChI

1S/C19H21N7O6/c20-19-25-15-14(17(30)26-19)23-11(8-22-15)7-21-10-3-1-9(2-4-10)16(29)24-12(18(31)32)5-6-13(27)28/h1-4,12,21H,5-8H2,(H,24,29)(H,27,28)(H,31,32)(H4,20,22,25,26,30)/t12-/m0/s1

InChI key

OZRNSSUDZOLUSN-LBPRGKRZSA-N

Gene Information

human ... DHFRP1(573971)
mouse ... Dhfr(13361)

Looking for similar products? Visit Product Comparison Guide

General description

Intermediate in mammalian conversion of dietary folic acid to tetrahydrofolate by dihydrofolate reductase (DHFR). In bacteria, dihydrofolic acid is generated from 7,8-dihydropteroate by dihydrofolate synthetase.

Application

Dihydrofolic acid has been used in diaphorase-coupled assay for dihydrofolate reductase (DHFR) activity. It has also been used as a component in assay buffer to determine dihydrofolate reductase (DHFR) activity.

Biochem/physiol Actions

Folic acid (FA) and dihydrofolic acid (FAH2) are substrates of dihydrofolate reductase(s) which reduce them to tetrahydrofolate (THF), which in turn supports ‘one carbon′ transfer. Tetrahydrofolates are required for de novo synthesis of purines, thymidylic acid and various amino acids and for post-translational methylation (epigenetics).

Packaging

Sealed ampule.

related product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James L Bodnar et al.
Ticks and tick-borne diseases, 9(3), 443-449 (2017-12-30)
Although nonpathogenic bacterial endosymbionts have been shown to contribute to their arthropod host's fitness by supplying them with essential vitamins and amino acids, little is known about the nutritional basis for the symbiotic relationship of endosymbionts in ticks. Our lab
David T Manallack
Journal of molecular modeling, 14(9), 797-805 (2008-05-27)
This study employed surface-based properties for use in the superimposition of three series of molecules. The properties used were derived from semiempirical molecular orbital calculations and can be related to the physics of intermolecular interactions. In each case, the superimposition
Z Attias et al.
Endocrine-related cancer, 13(2), 571-581 (2006-05-27)
The insulin-like growth factor-I receptor (IGF-IR) has an important role in colorectal cancer development and progression. IGF-IR displays a potent anti-apoptotic activity and is overexpressed in primary tumors and colon cancer-derived cell lines. Folic acid, a member of the vitamin
Karunesh Arora et al.
Journal of the American Chemical Society, 131(15), 5642-5647 (2009-03-28)
Conformational changes in enzymes are well recognized to play an important role in the organization of the reactive groups for efficient catalysis. This study reveals atomic and energetic details of the conformational change process that precedes the catalytic reaction of
Tobias Sahr et al.
The Biochemical journal, 396(1), 157-162 (2006-02-10)
pABA (p-aminobenzoate) is a precursor of folates and, besides esterification to glucose, has no other known metabolic fate in plants. It is synthesized in two steps from chorismate and glutamine, the first step being their conversion into glutamate and ADC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service