Skip to Content
Merck
All Photos(6)

Documents

U6751

Sigma-Aldrich

Uridine 5′-diphosphoglucuronic acid trisodium salt

98-100%

Synonym(s):

UDP-GlcA, UDPGA, Uridine-diphosphate-glucuronic acid trisodium salt, Uridine[5′]diphospho[1]-α-D-glucopyranosuronic acid trisodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H19N2Na3O18P2
CAS Number:
Molecular Weight:
646.23
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

bovine liver
rabbit muscle
yeast

Assay

98-100%

form

powder

storage temp.

−20°C

SMILES string

[Na+].[Na+].[Na+].O[C@@H]1[C@@H](O)[C@H](O[C@@H]([C@H]1O)C([O-])=O)OP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N3C=CC(=O)NC3=O

InChI

1S/C15H22N2O18P2.3Na/c18-5-1-2-17(15(26)16-5)12-9(22)6(19)4(32-12)3-31-36(27,28)35-37(29,30)34-14-10(23)7(20)8(21)11(33-14)13(24)25;;;/h1-2,4,6-12,14,19-23H,3H2,(H,24,25)(H,27,28)(H,29,30)(H,16,18,26);;;/q;3*+1/p-3/t4-,6-,7+,8+,9-,10-,11+,12-,14-;;;/m1.../s1

InChI key

XXXUNWUNTOMVIG-QWGSIYABSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Uridine 5′-diphosphoglucuronic acid trisodium salt has been used in optimization of incubation assay and glucuronidation assay.

Biochem/physiol Actions

Uridine 5′-diphosphate-glucuronic acid (UDP-GlcA) is a carboxylic acid-formed sugar nucleotide. It participates in the biosynthesis of numerous cell components. UDP-GlcA is a glycosyl donor for glycosyltransferases in the biosynthesis of several cell components like glycosaminoglycans, some N- and O-linked glycans, glycosphingolipids and pectins.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jan J Lyczakowski et al.
The New phytologist, 231(5), 1720-1733 (2021-06-05)
Wood of coniferous trees (softwood), is a globally significant carbon sink and an important source of biomass. Despite that, little is known about the genetic basis of softwood cell wall biosynthesis. Branching of xylan, one of the main hemicelluloses in
Comparing substrate specificity of two UDP-sugar pyrophosphorylases and efficient one-pot enzymatic synthesis of UDP-GlcA and UDP-GalA
Guo Y, et al.
Carbohydrate Research, 411, 1-5 (2015)
In vitro inhibitory effects of Andrographis paniculata, Gynura procumbens, Ficus deltoidea, and Curcuma xanthorrhiza extracts and constituents on human liver glucuronidation activity
Husni Z, et al.
Pharmacognosy magazine, 13( 2), S236-S236 (2017)
Soy isoflavone metabolism in cats compared with other species: urinary metabolite concentrations and glucuronidation by liver microsomes
Redmon J M, et al.
Xenobiotica, 46(5), 406-415 (2016)
Bao-Li Mi et al.
Xenobiotica; the fate of foreign compounds in biological systems, 44(8), 716-721 (2014-03-13)
1. The aurantio-obtusin's glucuronide was detected when aurantio-obtusin was incubated with human liver microsomes (HLMs). Recombinant UGT isoforms screening experiment showed that UGT1A8 was the major isoform contributed to the glucuronidation. 2. The metabolic profiles for aurantio-obtusin in liver microsomes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service