Skip to Content
Merck
All Photos(2)

Key Documents

91928

Supelco

2′,4′,6′-Trihydroxyacetophenone monohydrate

matrix substance for MALDI-MS, ≥99.5%

Synonym(s):

2-Acetylphloroglucinol, THAP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)3C6H2COCH3·H2O
CAS Number:
Molecular Weight:
186.16
Beilstein:
9288656
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21

grade

matrix substance for MALDI-MS

Quality Level

Assay

≥99.5% (HPLC)
≥99.5%

form

solid

analyte chemical class(es)

glycans, oligonucleotides, peptides, proteins

technique(s)

MALDI-MS: suitable

color

light yellow

mp

218-222 °C
219-221 °C (lit.)

cation traces

Ca: ≤200 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

suitability

in accordance for UV test
suitable for matrix substance for MALDI-MS

SMILES string

O.CC(=O)c1c(O)cc(O)cc1O

InChI

1S/C8H8O4.H2O/c1-4(9)8-6(11)2-5(10)3-7(8)12;/h2-3,10-12H,1H3;1H2

InChI key

GDSIBPPJKSBCMF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

MALDI-MS analyzes require highest purity matrices and reagents, as organic or trace metal contamination causes adverse effects during crystallization, ionization or measurement. Therefore, our portfolio of MALDI matrices and reagents such as 2′,4′,6′-Trihydroxyacetophenone monohydrate are designed to fulfill the demanding requirements of today′s challenging applications.

Application

2′,4′,6′-Trihydroxyacetophenone monohydrate has been used as a matrix for oligosaccharide and glycopeptide analysis using matrix-assisted laser desorption/ionization time-of-flight mass spectrometry (MALDI/TOF).
Used in MALDI analysis of acidic glycans and glyco­peptides in negative ion mode. Lower limit of detection than 6-aza-2-thiothimidine.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tao Wang et al.
The Analyst, 146(1), 289-295 (2020-11-04)
Although the in vivo metabolic pathways of scutellarin, a traditional Chinese medicine, have been investigated via different liquid chromatography techniques, studies on the distribution and location of scutellarin within organ tissue sections have not been reported. Matrix-assisted laser desorption/ionization mass
Analysis of acidic oligosaccharides and glycopeptides by matrix-assisted laser desorption/ionization time-of-flight mass spectrometry.
Papac, Damon I., Anissa Wong, and Andrew JS Jones.
Analytical Chemistry, 68.18, 3215-3223 (1996)
U Pieles et al.
Nucleic acids research, 21(14), 3191-3196 (1993-07-11)
We report the analysis and characterization of natural and modified oligonucleotides by matrix-assisted laser desorption ionization time-of-flight mass spectrometry (MALDI-TOF MS). The present technology was highly improved for this class of compounds by using a new matrix, 2,4,6-trihydroxy acetophenone, together
D I Papac et al.
Analytical chemistry, 68(18), 3215-3223 (1996-09-15)
2,5-Dihydroxybenzoic acid (DHB) is the most commonly used matrix for matrix-assisted laser desorption/ionization time-of-flight mass spectrometry (MALDI/TOF) of oligosaccharides. Because acidic, sialylated oligosaccharides are detected at only the low picomole level with DHB, alternative matrices were screened to identify a
J Wang et al.
Journal of agricultural and food chemistry, 47(5), 2009-2015 (1999-12-20)
Matrix-assisted laser desorption/ionization mass spectrometry (MALDI-MS) is a powerful new technique that will have a great impact on food analysis. This study is the first to demonstrate the applicability of MALDI-MS to perform both qualitative and quantitative analyses of anthocyanins

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service