Skip to Content
Merck
All Photos(1)

Key Documents

A0956

Sigma-Aldrich

Angelicin

Synonym(s):

2-Oxo-(2H)-furo(2,3-h)-1-benzopyran, 2H-Furo[2,3-h]-1-benzopyran-2-one, Isopsoralen, NSC 404563

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H6O3
CAS Number:
Molecular Weight:
186.16
Beilstein:
153970
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.25

form

powder

Quality Level

storage temp.

2-8°C

SMILES string

O=C1Oc2c(C=C1)ccc3occc23

InChI

1S/C11H6O3/c12-10-4-2-7-1-3-9-8(5-6-13-9)11(7)14-10/h1-6H

InChI key

XDROKJSWHURZGO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Angelicin, an angular furocoumarin, may be used as a photoactivatable molecule that reacts with DNA and unsaturated fatty acids to form monoadducts. Angelicin may be studied as a potential phototherapeutic and to understand its mechanisms of action and effects on RNA and DNA synthesis.

Biochem/physiol Actions

Angular furocoumarin with diverse photobiological effects. Upon long-wavelength UV irradiation, forms monoadduct with double-stranded DNA and reacts with unsaturated fatty acids. Inhibits DNA and RNA synthesis and cell replication in Ehrlich ascites tumor cells.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li-Qun Ouyang et al.
Analytica chimica acta, 650(2), 160-166 (2009-09-02)
A method using high performance liquid chromatography with photodiode-array detection (HPLC-DAD) coupled with alternating trilinear decomposition (ATLD) algorithm was proposed for simultaneous determination of psoralen and isopsoralen in plasma and Chinese medicine "Xian Ling Gu Bao" capsule (XLGBC). In this
Yang Ye et al.
Organic letters, 14(8), 2114-2117 (2012-04-07)
The oxidative dearomatization of 3-(3-alkynyl-4-hydroxyphenyl)propanoic acid is combined with a cascade transition-metal catalyzed cyclization/addition/aromatization/lactamization sequence, which provides a novel approach to prepare furoquinolinone and angelicin derivatives in a convergent and efficient manner.
Giampietro Viola et al.
Annals of the New York Academy of Sciences, 1171, 334-344 (2009-09-03)
In this review we summarize the structure and biological effects of linear and angular psoralens. These compounds exhibit interesting biological effects on the cell cycle, apoptosis and differentiation. These molecules should be considered promising drugs in the therapy of several
Liang Feng et al.
Archives of pharmacal research, 33(2), 225-230 (2010-03-03)
Coumarin components from Psoralea corylifolia L. are novel drugs in which psoralen and isopsoralen are the active components. The pharmacokinetics, tissue distribution and excretion of the two compounds were studied by liquid chromatography-tandem mass spectrometry after intravenous administration to Wistar
Yuan Gu et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 877(27), 3137-3143 (2009-08-13)
A rapid, specific and sensitive ultra-performance liquid chromatography/tandem mass spectrometry (UPLC/MS/MS) method has been established for simultaneous quantitation of psoralen and isopsoralen in rat plasma. Plasma samples were pretreated by direct protein precipitation with acetonitrile. Chromatographic separations were performed on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service