Skip to Content
Merck
All Photos(2)

Key Documents

69664

Supelco

Bergapten

analytical standard

Synonym(s):

5-Methoxypsoralen, 4-Methoxyfuro[3,2-g]benzopyrane-7-one, Bergapten, Geralen, Heraclin, Majudin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8O4
CAS Number:
Molecular Weight:
216.19
Beilstein:
19560
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

189-193 °C
190-193 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

COc1c2C=CC(=O)Oc2cc3occc13

InChI

1S/C12H8O4/c1-14-12-7-2-3-11(13)16-10(7)6-9-8(12)4-5-15-9/h2-6H,1H3

InChI key

BGEBZHIAGXMEMV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Muta. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Alessia Salvador et al.
ChemMedChem, 5(9), 1506-1512 (2010-07-21)
Induction of terminal erythroid differentiation can be an efficient strategy to inhibit proliferation of chronic myelogenous leukemia cells. Psoralens, well-known photo-chemotherapeutic agents, were found to be efficient at inducing erythroid differentiation of K562 cells, an in vitro cell line isolated
M Inzinger et al.
The British journal of dermatology, 165(3), 640-645 (2011-05-14)
Few studies have directly compared the clinical efficacy of psoralen plus ultraviolet A (PUVA) vs. biologics in the treatment of psoriasis. To compare the clinical efficacy of PUVA and biologic therapies for psoriasis under daily life conditions. Data from a
Sailesh Konda et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 49(5), 1092-1095 (2011-02-02)
5-methoxypsoralen (bergapten) has been used in the treatment of psoriasis and vitiligo, and as a sun tanning accelerator. While low plasma concentrations have previously been detected, there is no data on its dermatopharmacokinetics. Three rhesus monkeys were used as a
W McNeely et al.
Drugs, 56(4), 667-690 (1998-11-07)
5-Methoxypsoralen, a naturally occurring linear furocoumarin, has been successfully used in combination with ultraviolet (UV) A irradiation [psoralen plus UV (PUVA)] to manage psoriasis and vitiligo. In patients and volunteers, PUVA 5-methoxypsoralen causes a dose-related increase in cutaneous photosensitivity. However
Wei Yang et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 878(5-6), 575-582 (2010-02-02)
A sensitive, specific and rapid liquid chromatography-mass spectrometry (LC-MS) method has been developed and validated for the simultaneous determination of xanthotoxin (8-methoxypsoralen), psoralen, isoimpinellin (5,8-dimethoxypsoralen) and bergapten (5-methoxypsoralen) in rat plasma using pimpinellin as an internal standard (IS). The plasma

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service