Skip to Content
Merck
All Photos(1)

Key Documents

SML0552

Sigma-Aldrich

Tirapazamine

≥98% (HPLC)

Synonym(s):

4-Hydroxy-1-oxido-1,2,4-benzotriazin-1-ium-3-imine, SR 259075, SR 4233, Tirazone, Win 59075

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6N4O2
CAS Number:
Molecular Weight:
178.15
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

, orange to dark orange-red

solubility

DMSO: 10 mg/mL, clear

shipped in

wet ice

storage temp.

−20°C

SMILES string

Nc1n[n+]([O-])c2ccccc2[n+]1[O-]

InChI

1S/C7H6N4O2/c8-7-9-11(13)6-4-2-1-3-5(6)10(7)12/h1-4H,(H2,8,9)

InChI key

ORYDPOVDJJZGHQ-UHFFFAOYSA-N

Application

Tirapazamine has been used to evaluate its cytotoxic effect on U-251 MG (glioblastoma cell line) cell viability.

Biochem/physiol Actions

Under hypoxic conditions, tirapazamine is a potent cytotoxic agent that induces apoptosis by inducing breaks in single and double stranded DNA, as well as chromosomal breaks. The compound sensitizes cells to other ionizing radiation and other cytotoxic agents like cisplatin.
Under hypoxic conditions, tirapazamine is a potent cytotoxic agent..

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J M Brown
British journal of cancer, 67(6), 1163-1170 (1993-06-01)
SR 4233 (3-amino-1,2,4-benzotriazine 1,4-dioxide, WIN 59075, tirapazamine) is the lead compound in a new class of bioreductive anticancer drugs, the benzotriazine di-N-oxides. It is currently undergoing Phase I clinical testing. The preferential tumour cell killing of SR 4233 is a
Klaas M Govaert et al.
Annals of surgery, 259(4), 750-759 (2013-11-21)
To assess the contribution of hypoxia and bone marrow-derived cells to aggressive outgrowth of micrometastases after liver surgery. Liver surgery generates a microenvironment that fosters aggressive tumor recurrence. These areas are characterized by chronic hypoxia and influx of bone marrow-derived
W A Denny et al.
Expert opinion on investigational drugs, 9(12), 2889-2901 (2000-11-28)
Tirapazamine is the second clinical anticancer drug (after porfiromycin) that functions primarily as a hypoxia-selective cytotoxin. Hypoxic cells in tumours are relatively resistant to radiotherapy and to some forms of chemotherapy and are also biologically aggressive, thus representing an important
Yunzhou Shi et al.
Neoplasia (New York, N.Y.), 19(11), 950-959 (2017-10-11)
The effect of anti-angiogenic agents on tumor oxygenation has been in question for a number of years, where both increases and decreases in tumor pO
Yung-Ann Chen et al.
Lab on a chip, 11(21), 3626-3633 (2011-09-15)
This paper reports a microfluidic device capable of generating oxygen gradients for cell culture using spatially confined chemical reactions with minimal chemical consumption. The microfluidic cell culture device is constructed by single-layer polydimethylsiloxane (PDMS) microfluidic channels, in which the cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service