Skip to Content
Merck
All Photos(1)

Key Documents

A0298

Sigma-Aldrich

Adrenocorticotropic Hormone Fragment 1-24 human, rat

≥97% (HPLC)

Synonym(s):

Tetracosactide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C136H210N40O31S
CAS Number:
Molecular Weight:
2933.44
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.32
biological source:
synthetic (organic)
form:
powder

biological source

synthetic (organic)

Quality Level

sterility

non-sterile

Assay

≥97% (HPLC)

form

powder

composition

Peptide content, ≥60%

UniProt accession no.

storage temp.

−20°C

SMILES string

CSCC[C@H](NC(=O)[C@H](CO)NC(=O)[C@H](Cc1ccc(O)cc1)NC(=O)[C@@H](N)CO)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](Cc2c[nH]cn2)C(=O)N[C@@H](Cc3ccccc3)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](Cc4c[nH]c5ccccc45)C(=O)NCC(=O)N[C@@H](CCCCN)C(=O)N6CCC[C@H]6C(=O)N[C@@H](C(C)C)C(=O)NCC(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N7CCC[C@H]7C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](Cc8ccc(O)cc8)C(=O)N9CCC[C@H]9C(O)=O

InChI

1S/C136H210N40O31S/c1-75(2)109(127(200)154-71-106(181)156-88(31-13-17-52-137)114(187)158-89(32-14-18-53-138)115(188)159-91(35-21-56-149-134(142)143)116(189)164-96(37-23-58-151-136(146)147)131(204)175-60-25-39-104(175)126(199)173-111(77(5)6)128(201)163-90(33-15-19-54-139)120(193)171-110(76(3)4)129(202)169-101(65-80-43-47-84(180)48-44-80)132(205)176-61-26-40-105(176)133(206)207)172-125(198)103-38-24-59-174(103)130(203)95(34-16-20-55-140)157-107(182)70-153-113(186)99(66-81-68-152-87-30-12-11-29-85(81)87)167-117(190)92(36-22-57-150-135(144)145)160-121(194)98(63-78-27-9-8-10-28-78)166-123(196)100(67-82-69-148-74-155-82)168-118(191)93(49-50-108(183)184)161-119(192)94(51-62-208-7)162-124(197)102(73-178)170-122(195)97(165-112(185)86(141)72-177)64-79-41-45-83(179)46-42-79/h8-12,27-30,41-48,68-69,74-77,86,88-105,109-111,152,177-180H,13-26,31-40,49-67,70-73,137-141H2,1-7H3,(H,148,155)(H,153,186)(H,154,200)(H,156,181)(H,157,182)(H,158,187)(H,159,188)(H,160,194)(H,161,192)(H,162,197)(H,163,201)(H,164,189)(H,165,185)(H,166,196)(H,167,190)(H,168,191)(H,169,202)(H,170,195)(H,171,193)(H,172,198)(H,173,199)(H,183,184)(H,206,207)(H4,142,143,149)(H4,144,145,150)(H4,146,147,151)/t86-,88-,89-,90-,91-,92-,93-,94-,95-,96-,97-,98-,99-,100-,101-,102-,103-,104-,105-,109-,110-,111-/m0/s1

InChI key

ZOEFCCMDUURGSE-SQKVDDBVSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Ser-Tyr-Ser-Met-Glu-His-Phe-Arg-Trp-Gly-Lys-Pro-Val-Gly-Lys-Lys-Arg-Arg-Pro-Val-Lys-Val-Tyr-Pro

General description

Adrenocorticotropic hormone (ACTH), also called proopiomelanocortin (POMC), is a 303-amino acid hormone protein. Proteolysis of ACTH results in active peptides. ACTH is mapped to human chromosome 2p23.3.
The portion of the ACTH sequence that is conserved across species; has ~75% of the potency of ACTH on a molar basis.

Application

Adrenocorticotropic Hormone Fragment 1-24 human, rat, has been used in corticosterone enzyme immunoassay and to induce color change in frogs.

Biochem/physiol Actions

Activates G proteins.
Adrenocorticotropic hormone (ACTH) activates melanocortin receptor (MC1R to MC5R) and plays a key role in generation of α-melanocyte-stimulating hormone (α-MSH). The loss of function mutations in ACTH gene is implicated in obesity and red hair. Hypomethylation of POMC promoter region in thymic carcinoids is associated with ectopic adrenocorticotropic hormone syndrome. POMC mutation results in generation of abnormal fusion protein, impairing melanocortin signalling in hypoadrenalism. Dysregulation of ACTH gene transcription impacts metabolic pathways leading to Cushing disease.

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J K Jellyman et al.
Equine veterinary journal. Supplement, (41)(41), 38-42 (2012-05-19)
The effects of overexposure to glucocorticoids during early life of the foal on the subsequent HPA programming of the hypothalamic-pituitary-adrenal axis are unknown. To test the hypotheses that excess glucocorticoid exposure in early life subsequently increases both basal plasma concentrations
The effect of stress and stress hormones on dynamic colour-change in a sexually dichromatic Australian frog
Kindermann C, et al.
Comparative Biochemistry and Physiology. Part A, Molecular & Integrative Physiology, 165(2), 223-227 (2013)
O Hochwald et al.
Journal of perinatology : official journal of the California Perinatal Association, 32(6), 412-417 (2012-03-10)
To investigate if in preterm newborns, an early adrenocorticotropin hormone (ACTH) test can identify possible transient adrenal insufficiency (TAI), using significant hypotension as a clinical marker. We studied 40 premature newborns born 24 to 29 weeks gestational age (GA) before
Urinary corticosterone metabolites and chytridiomycosis disease prevalence in a free-living population of male Stony Creek frogs (Litoria wilcoxii)
Kindermann C, et al.
Comparative Biochemistry and Physiology. Part A, Molecular & Integrative Physiology, 162(3), 171-176 (2012)
Vijay K Yadav et al.
The Journal of clinical investigation, 132(4) (2021-12-15)
Through their ability to regulate gene expression in most organs, glucocorticoid (GC) hormones influence numerous physiological processes and are therefore key regulators of organismal homeostasis. In bone, GC hormones inhibit expression of the hormone Osteocalcin for poorly understood reasons. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service