Skip to Content
Merck
All Photos(1)

Documents

SML1527

Sigma-Aldrich

Abiraterone acetate

≥98% (HPLC)

Synonym(s):

CB 7598, 17-(Pyridin-3-yl)androsta-5,16-dien-3β-yl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H33NO2
CAS Number:
Molecular Weight:
391.55
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear

storage temp.

−20°C

SMILES string

C[C@]12C(C[C@@H](OC(C)=O)CC2)=CC[C@]3([H])[C@]1([H])CC[C@@]4(C)[C@@]3([H])CC=C4C5=CC=CN=C5

InChI

1S/C26H33NO2/c1-17(28)29-20-10-12-25(2)19(15-20)6-7-21-23-9-8-22(18-5-4-14-27-16-18)26(23,3)13-11-24(21)25/h4-6,8,14,16,20-21,23-24H,7,9-13,15H2,1-3H3/t20-,21-,23-,24-,25-,26+/m0/s1

InChI key

UVIQSJCZCSLXRZ-UBUQANBQSA-N

Gene Information

human ... CYP17A1(1586)

Looking for similar products? Visit Product Comparison Guide

Application

Abiraterone acetate has been used to study its antitumor action in 3D micro-tumour platform.

Biochem/physiol Actions

Abiraterone acetate is a prodrug of abiraterone, which is a potent, selective, and orally bioavailable inhibitor of CYP17A1 (CYP450c17), an enzyme that catalyzes two key serial reactions (17α hydroxylase and 17,20 lyase) in androgen and estrogen biosynthesis resulting in the formation of DHEA and androstenedione, which may ultimately be metabolized into testosterone. CYP17 is the key enzyme for androgen biosynthesis in both the testes and adrenals, so its inhibition should stop the production of androgens in both places. Abiraterone acetate is used for the treatment of metastatic castration-resistant prostate cancer. Abiraterone acetate possesses significant antitumor activity in post-docetaxel patients with CRPC (castration-resistant prostate cancer). It is highly essential for androgen biosynthesis in the testes, adrenal glands, and prostate tissue.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 1 - Repr. 1B - STOT RE 2

Target Organs

Endocrine system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gerhardt Attard et al.
Cancer research, 69(12), 4937-4940 (2009-06-11)
Abiraterone acetate is a potent, selective, and orally bioavailable small molecule inhibitor of CYP17, an enzyme that catalyzes two key serial reactions (17 alpha hydroxylase and 17,20 lyase) in androgen and estrogen biosynthesis. Clinical trials have confirmed that specific inhibition
Charles J Ryan et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 28(9), 1481-1488 (2010-02-18)
Abiraterone acetate is a prodrug of abiraterone, a selective inhibitor of CYP17, the enzyme catalyst for two essential steps in androgen biosynthesis. In castration-resistant prostate cancers (CRPCs), extragonadal androgen sources may sustain tumor growth despite a castrate environment. This phase
Alison H M Reid et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 28(9), 1489-1495 (2010-02-18)
The principal objective of this trial was to evaluate the antitumor activity of abiraterone acetate, an oral, specific, irreversible inhibitor of CYP17 in docetaxel-treated patients with castration-resistant prostate cancer (CRPC). In this multicenter, two-stage, phase II study, abiraterone acetate 1,000
E David Crawford et al.
The Journal of urology, 194(6), 1537-1547 (2015-07-22)
The availability of newly approved treatment options for metastatic castration resistant prostate cancer is not matched with conclusive data on optimal sequencing strategies and resistance patterns. A comprehensive review of efficacy and safety data for new agents and current knowledge
Daniel C Danila et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 28(9), 1496-1501 (2010-02-18)
Persistence of ligand-mediated androgen receptor signaling has been documented in castration-resistant prostate cancers (CRPCs). Abiraterone acetate (AA) is a potent and selective inhibitor of CYP17, which is required for androgen biosynthesis in the testes, adrenal glands, and prostate tissue. This

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service