Skip to Content
Merck
All Photos(1)

Key Documents

A0423

Sigma-Aldrich

Adrenocorticotropic Hormone human

≥97% (HPLC)

Synonym(s):

Adrenocorticotropic hormone fragment 1-39, Corticotropin A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C207H308N56O58S
CAS Number:
Molecular Weight:
4541.07
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.32
biological source:
synthetic (organic)
form:
powder
Assay:
≥97% (HPLC)
Pricing and availability is not currently available.

biological source

synthetic (organic)

Quality Level

sterility

non-sterile

Assay

≥97% (HPLC)

form

powder

solubility

water: 1.00-1.04 mg/mL, clear, colorless

UniProt accession no.

shipped in

ambient

storage temp.

−20°C

SMILES string

S(CCC(NC(=O)C(NC(=O)C(NC(=O)C(N)CO)Cc%12ccc(cc%12)O)CO)C(=O)NC(CCC(=O)O)C(=O)NC(Cc%11[nH]cnc%11)C(=O)NC(Cc%10ccccc%10)C(=O)NC(CCCN\C(=N\[H])\N)C(=O)NC(Cc8c9c([nH]c8)cccc9)C(=O)NCC(=O)NC(CCCCN)C(=O)N1C(CCC1)C(=O)NC(C(C)C)C(=O)NCC(=O)NC(CCCCN)C(=O)NC(CCCCN)

InChI

1S/C207H308N56O58S/c1-108(2)89-140(186(302)240-135(69-74-163(279)280)182(298)254-149(204(320)321)94-117-43-20-15-21-44-117)250-193(309)152-54-35-86-262(152)202(318)147(92-116-41-18-14-19-42-116)252-171(287)114(11)230-175(291)132(66-71-160(273)274)234-170(286)113(10)231-191(307)150(105-265)255-183(299)136(70-75-164(281)282)241-190(306)146(98-165(283)284)249-180(296)133(67-72-161(275)276)235-169(285)112(9)229-157(270)101-225-174(290)145(97-156(213)269)251-194(310)153-55-36-87-263(153)203(319)148(93-119-60-64-123(268)65-61-119)253-199(315)167(110(5)6)257-185(301)129(49-26-30-79-210)243-198(314)168(111(7)8)259-196(312)155-57-38-85-261(155)201(317)139(53-34-83-223-207(218)219)244-178(294)130(51-32-81-221-205(214)215)237-177(293)128(48-25-29-78-209)236-176(292)127(47-24-28-77-208)232-158(271)103-227-197(313)166(109(3)4)258-195(311)154-56-37-84-260(154)200(316)138(50-27-31-80-211)233-159(272)102-226-173(289)143(95-120-99-224-126-46-23-22-45-124(120)126)247-179(295)131(52-33-82-222-206(216)217)238-187(303)142(90-115-39-16-13-17-40-115)246-189(305)144(96-121-100-220-107-228-121)248-181(297)134(68-73-162(277)278)239-184(300)137(76-88-322-12)242-192(308)151(106-266)256-188(304)141(245-172(288)125(212)104-264)91-118-58-62-122(267)63-59-118/h13-23,39-46,58-65,99-100,107-114,125,127-155,166-168,224,264-268H,24-38,47-57,66-98,101-106,208-212H2,1-12H3,(H2,213,269)(H,220,228)(H,225,290)(H,226,289)(H,227,313)(H,229,270)(H,230,291)(H,231,307)(H,232,271)(H,233,272)(H,234,286)(H,235,285)(H,236,292)(H,237,293)(H,238,303)(H,239,300)(H,240,302)(H,241,306)(H,242,308)(H,243,314)(H,244,294)(H,245,288)(H,246,305)(H,247,295)(H,248,297)(H,249,296)(H,250,309)(H,251,310)(H,252,287)(H,253,315)(H,254,298)(H,255,299)(H,256,304)(H,257,301)(H,258,311)(H,259,312)(H,273,274)(H,275,276)(H,277,278)(H,279,280)(H,281,282)(H,283,284)(H,320,321)(H4,214,215,221)(H4,216,217,222)(H4,218,219,223)

InChI key

IDLFZVILOHSSID-UHFFFAOYSA-N

Gene Information

human ... POMC(5443)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Ser-Tyr-Ser-Met-Glu-His-Phe-Arg-Trp-Gly-Lys-Pro-Val-Gly-Lys-Lys-Arg-Arg-Pro-Val-Lys-Val-Tyr-Pro-Asn-Gly-Ala-Glu-Asp-Glu-Ser-Ala-Glu-Ala-Phe-Pro-Leu-Glu-Phe

General description

Adrenocorticotropic hormone (ACTH) is a 303-amino acid hormone protein,[1] which is located at human chromosome 2p23.3.[2]

Biochem/physiol Actions

Adrenocorticotropic hormone (ACTH) stimulates melanocortin receptor (MC1R to MC5R).It is implicated in the production of α-melanocyte-stimulating hormone (α-MSH).[1] Hypomethylation of ACTH promoter region in thymic carcinoids is associated with ectopic adrenocorticotropic hormone syndrome.[3] Mutations in this gene are associated with aberrant fusion protein and impairing melanocortin signaling in hypoadrenalism.[4] Dysregulation of ACTH leads to Cushing disease.[5]
Stimulates synthesis and secretion of glucocorticoids by adrenal cortex.

Other Notes

InChI: 1S/C207H308N56O58S/c1-108(2)89-140(186(302)240-135(69-74-163(279)280)182(298)254-149(204(320)321)94-117-43-20-15-21-44-117)250-193(309)152-54-35-86-262(152)202(318)147(92-116-41-18-14-19-42-116)252-171(287)114(11)230-175(291)132(66-71-160(273)274)234-170(286)113(10)231-191(307)150(105-265)255-183(299)136(70-75-164(281)282)241-190(306)146(98-165(283)284)249-180(296)133(67-72-161(275)276)235-169(285)112(9)229-157(270)101-225-174(290)145(97-156(213)269)251-194(310)153-55-36-87-263(153)203(319)148(93-119-60-64-123(268)65-61-119)253-199(315)167(110(5)6)257-185(301)129(49-26-30-79-210)243-198(314)168(111(7)8)259-196(312)155-57-38-85-261(155)201(317)139(53-34-83-223-207(218)219)244-178(294)130(51-32-81-221-205(214)215)237-177(293)128(48-25-29-78-209)236-176(292)127(47-24-28-77-208)232-158(271)103-227-197(313)166(109(3)4)258-195(311)154-56-37-84-260(154)200(316)138(50-27-31-80-211)233-159(272)102-226-173(289)143(95-120-99-224-126-46-23-22-45-124(120)126)247-179(295)131(52-33-82-222-206(216)217)238-187(303)142(90-115-39-16-13-17-40-115)246-189(305)144(96-121-100-220-107-228-121)248-181(297)134(68-73-162(277)278)239-184(300)137(76-88-322-12)242-192(308)151(106-266)256-188(304)141(245-172(288)125(212)104-264)91-118-58-62-122(267)63-59-118/h13-23,39-46,58-65,99-100,107-114,125,127-155,166-168,224,264-268H,24-38,47-57,66-98,101-106,208-212H2,1-12H3,(H2,213,269)(H,220,228)(H,225,290)(H,226,289)(H,227,313)(H,229,270)(H,230,291)(H,231,307)(H,232,271)(H,233,272)(H,234,286)(H,235,285)(H,236,292)(H,237,293)(H,238,303)(H,239,300)(H,240,302)(H,241,306)(H,242,308)(H,243,314)(H,244,294)(H,245,288)(H,246,305)(H,247,295)(H,248,297)(H,249,296)(H,250,309)(H,251,310)(H,252,287)(H,253,315)(H,254,298)(H,255,299)(H,256,304)(H,257,301)(H,258,311)(H,259,312)(H,273,274)(H,275,276)(H,277,278)(H,279,280)(H,281,282)(H,283,284)(H,320,321)(H4,214,215,221)(H4,216,217,222)(H4,218,219,223)/t112-,113-,114-,125-,127-,128-,129-,130-,131-,132-,133-,134-,135-,136-,137-,138-,139-,140-,141-,142-,143-,144-,145-,146-,147-,148-,149-,150-,151-,152-,153-,154-,155-,166-,167-,168-/m0/s1
Lyophilized from 0.1% TFA in H2O

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Athanasios Samaras et al.
Animals : an open access journal from MDPI, 12(24) (2022-12-24)
Cortisol concentration in fish scales is a novel and reliable indicator of chronic stress. However, until now cortisol in scales has been considered to be accumulated through the circulation and it has not yet been studied whether it can be
Adrenocorticotropins. 44. Total synthesis of the human hormone by the solid-phase method.
D Yamashiro et al.
Journal of the American Chemical Society, 95(4), 1310-1315 (1973-02-21)
Unexpected endocrine features and normal pigmentation in a young adult patient carrying a novel homozygous mutation in the POMC gene
Clement K, et al.
The Journal of Clinical Endocrinology and Metabolism, 93(12), 4955-4962 (2008)
Role of Brg1 and HDAC2 in GR trans-repression of the pituitary POMC gene and misexpression in Cushing disease
Bilodeau S, et al.
Genes & Development, 20(20), 2871-2886 (2006)
Sandra Sigala et al.
Cells, 11(9) (2022-05-15)
Adrenocortical carcinoma is a heterogeneous and aggressive cancer that originates from steroidogenic cells within the adrenal cortex. In this study, we have assessed for the preclinical gold standard NCI-H295 in direct comparison with the more recently established MUC-1 and a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service