Skip to Content
Merck
All Photos(1)

Key Documents

95126

Sigma-Aldrich

Triethylene glycol

BioUltra, anhydrous, ≥99.0% (GC)

Synonym(s):

Triglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2CH2O)2CH2CH2OH
CAS Number:
Molecular Weight:
150.17
Beilstein:
969357
EC Number:
MDL number:
UNSPSC Code:
12352136
PubChem Substance ID:
NACRES:
NA.25
technique(s):
GC/GC: suitable
bp:
125-127 °C/0.1 mmHg (lit.)
vapor pressure:
<0.01 mmHg ( 20 °C)
Pricing and availability is not currently available.

vapor density

5.2 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)

product line

BioUltra

Assay

≥99.0% (GC)

form

viscous liquid

autoignition temp.

699 °F

expl. lim.

9.2 %

technique(s)

GC/GC: suitable

impurities

insoluble matter, passes filter test
≤0.01% peroxides (as H2O2)

refractive index

n20/D 1.455 (lit.)
n20/D 1.456

pH

5.5-7.0 (25 °C, 50 mg/mL in H2O)

bp

125-127 °C/0.1 mmHg (lit.)

mp

−7 °C (lit.)

solubility

H2O: 50 mg/mL at 20 °C, clear, colorless

density

1.124 g/mL at 20 °C (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤200 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤200 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

OCCOCCOCCO

λ

50 mg/mL in H2O

UV absorption

λ: 260 nm Amax: ≤0.10
λ: 280 nm Amax: ≤0.05

InChI

1S/C6H14O4/c7-1-3-9-5-6-10-4-2-8/h7-8H,1-6H2

InChI key

ZIBGPFATKBEMQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Triethylene glycol may be used in nanotechnology as a functionalization reagent.

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

330.8 °F - closed cup

Flash Point(C)

166 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naoto Koizumi et al.
Journal of synchrotron radiation, 19(Pt 1), 74-83 (2011-12-22)
In situ Co K-edge quick-EXAFS (QEXAFS) coupled with temperature-programmed oxidation as well as ex situ XAFS was applied to investigating the mechanism for enhancing the dispersion of Co(3)O(4) nanoparticles in a calcined Co/SiO(2) Fischer-Tropsch synthesis catalyst prepared by adding triethylene
G Marchegiani et al.
Ultrasonics sonochemistry, 19(4), 877-882 (2012-01-13)
Hydrophilic magnetite nanoparticles in the size range 30-10nm are easily and rapidly prepared under ultrasonic irradiation of Fe(OH)(2) in di- and tri-ethylene glycol/water solution with volume ratio varying between 7:3 and 3:7. Structural (XRD) and morphological (SEM) characterization reveal good
Zi-Fu Wang et al.
Nucleic acids research, 40(17), 8711-8720 (2012-06-28)
Because various non-parallel G-quadruplexes of human telomeric sequences in K+ solution can be converted to a parallel G-quadruplex by adding polyethylene glycol (PEG) as a co-solvent, we have taken advantage of this property of PEG to study the covalent attachment
Maria I Montanez et al.
Biomacromolecules, 12(6), 2114-2125 (2011-04-29)
Well-defined dendronized cellulose substrates displaying multiple representations of dual-functionality were constructed by taking advantage of the efficiency of the click reaction combined with traditional anhydride chemistry. First, activated cellulose surfaces were decorated with several generations of dendrons, and their peripheral
Lei Dong et al.
Nanoscale, 3(9), 3447-3461 (2011-07-26)
A p/n heterojunction is the basic setup for light-electric conversion. It has been widely accepted that the ideal configuration for organic photovoltaics is formed by the joint of a pair of long-range continuous but nanometre-wide phases consisting of electron-donating (D)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service