Skip to Content
Merck
All Photos(1)

Key Documents

32160

Sigma-Aldrich

Diethylene glycol

puriss. p.a., ≥99.0% (GC), colorless

Synonym(s):

2,2′-Oxydiethanol, 2-Hydroxyethyl ether, Bis(2-hydroxyethyl) ether, Diglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HOCH2CH2)2O
CAS Number:
Molecular Weight:
106.12
Beilstein:
969209
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.0% (GC)
bp:
245 °C (lit.)
vapor pressure:
0.01 mmHg ( 20 °C)

vapor density

2.14 (vs air)

Quality Level

vapor pressure

0.01 mmHg ( 20 °C)

description

non-ionic

grade

puriss. p.a.

Assay

≥99.0% (GC)

form

viscous liquid (clear)

autoignition temp.

442 °F

expl. lim.

2-12.3 %

impurities

≤0.003% peroxides (as H2O2)
≤0.01% free acid (as CH3COOH)
≤0.1% water
≤0.8% ethylene glycol (HOCH2CH2OH)

color

colorless
APHA: ≤25

refractive index

n20/D 1.447 (lit.)
n20/D 1.447

bp

245 °C (lit.)

mp

−10 °C (lit.)

density

1.116 g/mL at 20 °C
1.118 g/mL at 25 °C (lit.)

cation traces

Ca: ≤5 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤2 mg/kg
K: ≤20 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Na: ≤20 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Zn: ≤10 mg/kg

SMILES string

OCCOCCO

InChI

1S/C4H10O3/c5-1-3-7-4-2-6/h5-6H,1-4H2

InChI key

MTHSVFCYNBDYFN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethylene glycol (DEG) is formed as side reaction product during the esterification reaction of terephthalic acid with ethylene glycol. It readily forms complexes with 3d transition metal cations in the presence of alkaline hydroxide. Its solutions are widely employed for antifreeze and lubricating applications. Oxidative degradation of DEG with ozone by modified Fenton process (mixture of hydrogen peroxide and ferric salt) was described in a study.

Application

Diethylene glycol may be used for electrodeposition of cadmium sulfide (CdSe) films. It may be used to compose the fluoride ion electrolytes, for use in the preparation of self-organized titania nanotube arrays.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

280.4 °F - closed cup

Flash Point(C)

138 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of ordered arrays of discrete, partially crystalline titania nanotubes by Ti anodization using diethylene glycol electrolytes.
Yoriya S, et al.
Journal of Materials Chemistry, 18(28), 3332-3336 (2008)
Daniela Caruntu et al.
Inorganic chemistry, 41(23), 6137-6146 (2002-11-12)
Study of the reactivity of 3d transition metal cations in diethylene glycol solutions revealed several key features that made it possible to develop a new method for synthesis of the nanocrystalline transition metal ferrites. The 3-7 nm particles of [MFe2O4]n[O2CR]m
Formation of diethylene glycol as a side reaction during production of polyethylene terephthalate.
Hovenkamp SG and Munting JP.
Journal of Polymer Science Part A: Polymer Chemistry, 8(3), 679-682 (1970)
The physical properties of CdS thin films electrodeposited from aqueous diethylene glycol solutions.
Baranski AS, et al.
Journal of Applied Physics, 54(11), 6390-6394 (1983)
Tuba Turan-Ertas et al.
Chemosphere, 47(3), 293-301 (2002-05-09)
This paper describes a study of oxidation of diethylene glycol (DEG) by ozone and modified Fenton process (hydrogen peroxide and ferric salt mixture) in aqueous solution. Both oxidation processes were able to oxidize relatively high concentrations of DEG effectively. DEG

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service