Skip to Content
Merck
All Photos(1)

Key Documents

D6001

Sigma-Aldrich

Dextran sulfate sodium salt from Leuconostoc spp.

average mol wt >500,000 (dextran starting material), contains 0.5-2.0% phosphate buffer, pH 6-8

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.21

biological source

bacterial (Leuconostoc mesenteroides)

Quality Level

form

powder

optical activity

[α]/D 90 to 105 °, c = 6% (w/v) in water

mol wt

average mol wt >500,000 (dextran starting material)

contains

0.5-2.0% phosphate buffer, pH 6-8

composition

Sulfur Content, 16.0-19.0%

color

white to off-white

useful pH range

6.0-8.0 (1% in water)

solubility

H2O: 100 mg/mL

Looking for similar products? Visit Product Comparison Guide

General description

Dextran sulfate is a negatively charged sulfated polysaccharide, which is produced by the esterification of dextran with chlorosulphonic acid. It has antiviral, anticoagulant and adjuvant properties. Dextran is used to treat hemorrhage, burns, surgery and trauma. It enhances blood flow and blocks erythrocyte aggregation. Dextran decreases the risk of thrombosis and postoperative pulmonary emboli. It acts as a ribonuclease inhibitor.

Application

Dextran sulfate sodium salt Leuconostoc spp has been used:
  • to delipidate fatty plasma(67)(70)
  • to induce inflammatory and carcinogenic response in the distal colon of rodents(68)
  • for the preparation of 3,4-ethylenedioxythiophene (EDOT)(69)

Other Notes

To gain a comprehensive understanding of our extensive range of Dextrans for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zora Djuric et al.
The Journal of nutritional biochemistry, 46, 90-99 (2017-05-10)
Dietary fish oils have potential for prevention of colon cancer, and yet the mechanisms of action in normal and tumor colon tissues are not well defined. Here we evaluated the impact of the colonic fatty acid milieu on the formation
Seda Onder et al.
Frontiers in pharmacology, 8, 713-713 (2017-10-27)
Hupresin is a new affinity resin that binds butyrylcholinesterase (BChE) in human plasma and acetylcholinesterase (AChE) solubilized from red blood cells (RBC). Hupresin is available from the CHEMFORASE company. BChE in human plasma binds to Hupresin and is released with
The Molecular Biology of Poliovirus (2012)
Rheology of Industrial Polysaccharides: Theory and Applications (2012)
Radiotherapy, Surgery, and Immunotherapy (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service