Skip to Content
Merck
All Photos(1)

Key Documents

88071

Sigma-Aldrich

meso-Tetraphenylporphyrin

BioReagent, suitable for fluorescence, ≥99.0% (HPLC)

Synonym(s):

5,10,15,20-Tetraphenylporphin, TPP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H30N4
CAS Number:
Molecular Weight:
614.74
Beilstein:
379542
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

Assay

≥99.0% (HPLC)

form

crystals

impurities

≤0.5% chlorin

solubility

dichloromethane: soluble

fluorescence

λex 420 nm; λem 651 nm in toluene

suitability

suitable for fluorescence

storage temp.

−20°C

SMILES string

c1ccc(cc1)-c2c3ccc(n3)c(-c4ccccc4)c5ccc([nH]5)c(-c6ccccc6)c7ccc(n7)c(-c8ccccc8)c9ccc2[nH]9

InChI

1S/C44H30N4/c1-5-13-29(14-6-1)41-33-21-23-35(45-33)42(30-15-7-2-8-16-30)37-25-27-39(47-37)44(32-19-11-4-12-20-32)40-28-26-38(48-40)43(31-17-9-3-10-18-31)36-24-22-34(41)46-36/h1-28,45,48H/b41-33-,41-34-,42-35-,42-37-,43-36-,43-38-,44-39-,44-40-

InChI key

YNHJECZULSZAQK-LWQDQPMZSA-N

Gene Information

human ... TERT(7015)

Looking for similar products? Visit Product Comparison Guide

General description

Tetraphenylporphyrin is a precipitation inhibitor.

Biochem/physiol Actions

Tetraphenylporphyrin serves as an efficient inhibitor for Mg and its alloys. It is mainly used as a porphyrin moiety, in the preparation of ligand and introduction of metal in porphyrin.

Analysis Note

In addition to the emission maximum at 651 nm, there is a lower maxima at 717 nm.

Other Notes

Sensitizer used for the photochemical generation of singlet oxygen

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E.D. Mihelich et al.
The Journal of Organic Chemistry, 48, 4135-4135 (1983)
Andrea Pantoja et al.
The Journal of infectious diseases, 211 Suppl 2, S67-S77 (2015-03-15)
Four priority target product profiles for the development of diagnostic tests for tuberculosis were identified: 1) Rapid sputum-based (RSP), 2) non-sputum Biomarker-based (BMT), 3) triage test followed by confirmatory test (TT), and 4) drug-susceptibility testing (DST). We assessed the cost
Chang Yin et al.
Frontiers in pharmacology, 8, 726-726 (2017-11-09)
Stem cells display a fundamentally different mechanism of proliferation control when compared to somatic cells. Uncovering these mechanisms would maximize the impact in drug discovery with a higher translational applicability. The unbiased approach used in phenotype-based drug discovery (PDD) programs
Ken-ichiro Murata et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(19), 5956-5961 (2015-04-29)
A liquid-liquid transition (LLT) in a single-component substance is an unconventional phase transition from one liquid to another. LLT has recently attracted considerable attention because of its fundamental importance in our understanding of the liquid state. To access the order
Xavier C Ding et al.
PLoS neglected tropical diseases, 11(4), e0005516-e0005516 (2017-04-04)
The global prevalence of malaria has decreased over the past fifteen years, but similar gains have not been realized against Plasmodium vivax because this species is less responsive to conventional malaria control interventions aimed principally at P. falciparum. Approximately half

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service