Skip to Content
Merck
All Photos(1)

Key Documents

252204

Sigma-Aldrich

5,10,15,20-Tetraphenyl-21H,23H-porphine nickel(II)

dye content ≥95 %

Synonym(s):

Nickel TPP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H28N4Ni
CAS Number:
Molecular Weight:
671.41
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

form

solid

composition

dye content, ≥95%

reaction suitability

reagent type: catalyst
core: nickel

λmax

525 nm (2nd)

SMILES string

[Ni]1n2c3ccc2c(-c4ccccc4)c5ccc(n5)c(-c6ccccc6)c7ccc(c(-c8ccccc8)c9ccc(n9)c3-c%10ccccc%10)n17

InChI

1S/C44H28N4.Ni/c1-5-13-29(14-6-1)41-33-21-23-35(45-33)42(30-15-7-2-8-16-30)37-25-27-39(47-37)44(32-19-11-4-12-20-32)40-28-26-38(48-40)43(31-17-9-3-10-18-31)36-24-22-34(41)46-36;/h1-28H;/q-2;+2/b41-33-,41-34-,42-35-,42-37-,43-36-,43-38-,44-39-,44-40-;

InChI key

CXIRWLOIAQYBLZ-DAJBKUBHSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qing Huang et al.
The journal of physical chemistry. A, 109(46), 10493-10502 (2006-07-13)
We have correlated the Raman intensities of out-of-plane modes of nickel porphyrins with the nonplanar deformations of specific symmetries, i.e., static normal coordinate deformations (SNCDs) expressed in terms of irreducible representations of the unperturbed D(4h) point group. The model porphyrins

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service