Skip to Content
Merck
All Photos(1)

Key Documents

84110

Sigma-Aldrich

Sucrose monolaurate

BioXtra, ≥97.0% (TLC)

Synonym(s):

n-Dodecanoylsucrose, Lauric acid sucrose ester, Sucrose monododecanoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H44O12
CAS Number:
Molecular Weight:
524.60
Beilstein:
65819
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

description

non-ionic

Quality Level

product line

BioXtra

Assay

≥97.0% (TLC)

form

powder

mol wt

524.60 g/mol

impurities

≤3% water

CMC

0.3

cation traces

Ca: ≤50 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤1000 mg/kg
Mg: ≤10 mg/kg
Mn: ≤5 mg/kg
Na: ≤500 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤10 mg/kg

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCC(=O)OC[C@H]1O[C@H](O[C@]2(CO)O[C@H](CO)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C24H44O12/c1-2-3-4-5-6-7-8-9-10-11-17(27)33-13-16-18(28)20(30)21(31)23(34-16)36-24(14-26)22(32)19(29)15(12-25)35-24/h15-16,18-23,25-26,28-32H,2-14H2,1H3/t15-,16-,18-,19-,20+,21-,22+,23-,24+/m1/s1

InChI key

KGUHOFWIXKIURA-VQXBOQCVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sucrose monolaurate is a non-ionic detergent and a carbohydrate fatty acid monoester. It is generally used for the solubilization of membrane proteins.[1]

Application

Sucrose monolaurate has been used as an antimicrobial agent.[2] It has also been used as a constituent of lysis buffer to homogenize placental tissues for determining serine palmitoyltransferase (SPT) activity.[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Detergents for extraction of visual pigments: types, solubilization, and stability.
S L Fong et al.
Methods in enzymology, 81, 133-140 (1982-01-01)
Prediction of Listeria monocytogenes ATCC 7644 growth on fresh-cut produce treated with bacteriophage and sucrose monolaurate by using artificial neural network
Oladunjoye A, et al.
LWT--Food Science and Technology, 76, 9-17 (2017)
Christopher Rupp et al.
International journal of pharmaceutics, 395(1-2), 272-280 (2010-06-29)
A remarkable part of newly developed active pharmaceutical ingredients is rejected in early phase development and will never find a way to a patient because of poor water solubility which is often paired with poor bioavailability. Considering such arising solubility
Mitsugu Shimobayashi et al.
Molecular biology of the cell, 24(6), 870-881 (2013-02-01)
The evolutionarily conserved Orm1 and Orm2 proteins mediate sphingolipid homeostasis. However, the homologous Orm proteins and the signaling pathways modulating their phosphorylation and function are incompletely characterized. Here we demonstrate that inhibition of nutrient-sensitive target of rapamycin complex 1 (TORC1)
Angéla Szuts et al.
International journal of pharmaceutics, 410(1-2), 107-110 (2011-03-23)
This study focused on an investigation of the applicability of sucrose laurate as surfactant in solid dispersions. Although this surfactant has a US Drug Master File, it has not been used so far in internal pharmaceutical products. High drug-loaded solid

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service