Skip to Content
Merck

Skip To

C4151

Sodium decanoate

≥98%

Synonym(s):

Capric acid sodium salt, Decanoic acid sodium salt, Sodium caprate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
CH3(CH2)8COONa
CAS Number:
Molecular Weight:
194.25
UNSPSC Code:
12352211
NACRES:
NA.25
PubChem Substance ID:
EC Number:
213-688-4
Beilstein/REAXYS Number:
3572742
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

natural (organic)

Quality Segment

assay

≥98%

form

powder

functional group

ester

lipid type

saturated FAs

shipped in

ambient

storage temp.

2-8°C

SMILES string

[Na+].CCCCCCCCCC([O-])=O

InChI

1S/C10H20O2.Na/c1-2-3-4-5-6-7-8-9-10(11)12;/h2-9H2,1H3,(H,11,12);/q;+1/p-1

InChI key

FIWQZURFGYXCEO-UHFFFAOYSA-M

Application


  • Neonatal intestinal mucus barrier changes in response to maturity, inflammation, and sodium decanoate supplementation.: This study elucidates how sodium decanoate affects neonatal intestinal barriers, indicating its potential for nutritional interventions to enhance mucosal immunity and gastrointestinal health (Ronholt et al., 2024).

  • Effects of Sodium Salts of Fatty Acids and Their Derivatives on Skin Permeation of Cromolyn Sodium.: Demonstrates the role of sodium decanoate in enhancing skin permeability, offering insights into its applications in transdermal drug delivery systems (Dinh et al., 2023).

  • The Role of Paracellular Transport in the Intestinal Absorption and Biopharmaceutical Characterization of Minoxidil.: Highlights sodium decanoate′s impact on the intestinal absorption mechanisms, contributing to the development of more effective oral drug formulations (Dahan et al., 2022).

  • Control of Escherichia coli Serotype O157:H7 Motility and Biofilm Formation by Salicylate and Decanoate: MarA/SoxS/Rob and pchE Interactions.: Investigates how sodium decanoate influences bacterial behavior, particularly in managing pathogenicity and biofilm control, which is crucial for both clinical and environmental applications (Ream et al., 2022).

  • Sodium Decanoate Improves Intestinal Epithelial Barrier and Antioxidation via Activating G Protein-Coupled Receptor-43.: This research explores sodium decanoate′s therapeutic potential in strengthening intestinal barriers and enhancing antioxidative properties, making it a candidate for gastroenterological treatments (Ma et al., 2021).


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
L9755S3381O7501
biological source

natural (organic)

biological source

synthetic

biological source

plant (vegetable)

biological source

synthetic (organic)

assay

≥98%

assay

99-100%

assay

≥99%

assay

≥99%

functional group

ester

functional group

ester

functional group

ester

functional group

ester

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient

form

powder

form

powder

form

powder

form

powder

storage temp.

2-8°C

storage temp.

room temp

storage temp.

2-8°C

storage temp.

−20°C


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Is C4151 reference material? I saw that for Sodium Caprylate there is a specific reference material that can be ordered but not for caprate?

    1 answer
    1. No, this product is not a certified reference material.

      Helpful?

Reviews

No rating value

Active Filters