Skip to Content
Merck
All Photos(1)

Key Documents

83895

Sigma-Aldrich

D-Ribulose 1,5-bisphosphate sodium salt hydrate

≥99.0% (TLC)

Synonym(s):

D-erythro-2-Pentulose, 1,5-bis(dihydrogen phosphate), D-Ribulose 1,5-diphosphate, RuDP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12O11P2 · xNa+ · yH2O
CAS Number:
Molecular Weight:
310.09 (anhydrous free acid basis)
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

Assay

≥99.0% (TLC)

form

powder

color

white

solubility

H2O: 50 mg/mL, clear, colorless to light yellow

storage temp.

−20°C

SMILES string

[P](=O)(OC[C@@H](O)[C@@H](O)C(=O)CO[P](=O)(O)O)(O)O

InChI

1S/C5H12O11P2/c6-3(1-15-17(9,10)11)5(8)4(7)2-16-18(12,13)14/h3,5-6,8H,1-2H2,(H2,9,10,11)(H2,12,13,14)/t3-,5-/m1/s1

InChI key

YAHZABJORDUQGO-NQXXGFSBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ribulose-1,5-bisphosphate (RuBP) is a component of the Calvin cycle that is metabolized into glycerate 3-phosphate (G3P) by the enzyme ribulose bisphosphate carboxylase/oxygenase (RuBisCO). RuBP is used to identify, differentiate and characterize ribulose bisphosphate carboxylase(s)/oxygenase(s) (RuBisCO).

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.
Active site studies with ribulose-1,5-biphosphate carboxylase

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Ogushi et al.
The Journal of biological chemistry, 265(19), 10943-10949 (1990-07-05)
The tissue contents of previously known allosteric effectors of brain phosphofructokinase (EC 2.7.1.11) (PFK) and the kinetic behavior of isolated PFK were investigated during the initiation of rapid glycolytic flux in freeze-blown rat brain. Comparing 0- with 5-s brains revealed
Takaaki Sato et al.
Science (New York, N.Y.), 315(5814), 1003-1006 (2007-02-17)
The type III ribulose-1,5-bisphosphate carboxylase-oxygenase (RuBisCO) present in the archaeon Thermococcus kodakaraensis was found to participate in adenosine 5'-monophosphate (AMP) metabolism, a role that is distinct from that of classical RuBisCOs of the Calvin-Benson-Bassham cycle. Genes annotated as thymidine phosphorylase
T Kawaguchi et al.
The Journal of biological chemistry, 276(30), 28554-28561 (2001-05-25)
Macrophages can adapt to the absence of oxygen by switching to anaerobic glycolysis. In this study, we investigated (a) the roles of fructose 2,6-bisphosphate (Fru-2,6-P2) and ribose 1,5-bisphosphate (Rib-1,5-P2), potent activators of phosphofructokinase, (b) the enzymes responsible for the synthesis
I Ozaki et al.
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology, 125(1), 97-102 (2000-06-07)
Fructose-1,6-bisphosphatase is one of the regulatory enzymes of gluconeogenesis in kidney cortex. The effect of ribose 1,5-bisphosphate on fructose-1,6-bisphosphatase purified from rat kidney cortex was studied. Rat kidney cortex, fructose-1,6-bisphosphatase exhibited hyperbolic kinetics with regard to its substrate, but the
M Willemoës et al.
Biochemistry, 36(16), 5078-5083 (1997-04-22)
The mechanism of binding of the substrates Mg x ATP and ribose 5-phosphate as well as Mg2+ to the enzyme 5-phospho-D-ribosyl (alpha-1-diphosphate synthetase from Escherichia coli has been analyzed. By use of the competive inhibitors of ATP and ribose 5-phosphate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service