83899
D-Ribulose 5-phosphate disodium salt
≥96% (TLC)
Synonym(s):
ᴅ-Ribulose 5-phosphate Disodium
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C5H9Na2O8P
CAS Number:
Molecular Weight:
274.07
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25
Recommended Products
Quality Level
Assay
≥96% (TLC)
form
powder
impurities
≤1% methanol
≤2% ethanol
<10% water
color
white to light beige
solubility
H2O: 50 mg/mL, clear, light yellow
storage temp.
−20°C
SMILES string
[Na+].[Na+].OCC(=O)[C@H](O)[C@H](O)COP([O-])([O-])=O
InChI
1S/C5H11O8P.2Na/c6-1-3(7)5(9)4(8)2-13-14(10,11)12;;/h4-6,8-9H,1-2H2,(H2,10,11,12);;/q;2*+1/p-2/t4-,5+;;/m1../s1
InChI key
NBEPTNUBEFMKAU-CIFXRNLBSA-L
Application
Ribulose 5-phosphate, a component of both the pentose phosphate pathway (PPP) and the Calvin cycle, is used as a substrate to identify, differentiate and characterize phosphopentose isomerase(s) and epimerase(s).
Other Notes
To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service