Skip to Content
Merck
All Photos(3)

Documents

D7137

Sigma-Aldrich

Dihydroxyacetone phosphate dilithium salt

≥93% dry basis (enzymatic)

Synonym(s):

1,3-Dihydroxy-2-propanone 1-phosphate dilithium salt, 1-Hydroxy-3-(phosphonooxy)-2-propanone dilithium salt, DHAP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5Li2O6P
CAS Number:
Molecular Weight:
181.92
Beilstein:
1708891
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

bovine (casein)

Quality Level

Assay

≥93% dry basis (enzymatic)

form

powder

impurities

~0.2 mol % D-glyceraldehyde 3-phosphate

color

white

solubility

soluble 50mg plus 1ml of H2O, clear, colorless to faintly yellow

anion traces

phosphate (PO43-): ~5 mol %

cation traces

Li: 5.5-8.0% (dry basis)

storage temp.

−20°C

SMILES string

[Li+].[Li+].OCC(=O)COP([O-])([O-])=O

InChI

1S/C3H7O6P.2Li/c4-1-3(5)2-9-10(6,7)8;;/h4H,1-2H2,(H2,6,7,8);;/q;2*+1/p-2

InChI key

QWIKESRFRWLYIA-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Dihydroxyacetone phosphate (DHAP) is used in fructose metabolism, triose and glycation research. It is used in studies involving the metabolism of three-carbon sugar pools and their roles in physiological and physical processes. DHAP may be used as a substrate to help identify, differentiate and characterize fructose-bisphosphate aldolase(s) (ALDOA) and triosephosphate isomerase(s). DHAP may be used to study cellular glycation stress.

Biochem/physiol Actions

Dihydroxyacetone phosphate (DHAP) is a metabolic intermediate involved in many pathways, including glycolysis, gluconeogenesis, glycerol metabolism, phosphatidic acid synthesis, fat metabolism, and the Calvin cycle.

Caution

Unlike the dimethyl ketal, does not require hydrolysis prior to use as a substrate.

Preparation Note

Enzymatically prepared.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chiara Fania et al.
PloS one, 12(6), e0179280-e0179280 (2017-06-20)
In the diagnosis of Alzheimer's disease (AD) total tau (T-tau), tau phosphorylated at threonine 181 (P-tau181), and the 42 amino acid isoform of alpha β-amyloid (Aβ) are well established surrogate CSF markers. However, there is a constant need for new
Saheem Ahmad et al.
PloS one, 8(9), e72128-e72128 (2013-09-12)
Non-enzymatic glycation is the addition of free carbonyl group of reducing sugar to the free amino groups of proteins, resulting in the formation of a Schiff base and an Amadori product. Dihydroxyacetone (DHA) is one of the carbonyl species which
Xi Luo et al.
Scientific reports, 7(1), 15170-15170 (2017-11-11)
Matrix-assisted laser desorption/ionization (MALDI) mass spectrometry has been widely used for biomolecular analysis. However, with conventional MALDI, it is difficult to analyse low-molecular-weight compounds because of the interference of matrix ion signals. Here, we report a matrix-free on-chip pulse-heating desorption/ionization
Daisuke Takagi et al.
Plant & cell physiology, 55(2), 333-340 (2014-01-11)
Sugar-derived reactive carbonyls (RCs), including methylglyoxal (MG), are aggressive by-products of oxidative stress known to impair the functions of multiple proteins. These advanced glycation end-products accumulate in patients with diabetes mellitus and cause major complications, including arteriosclerosis and cardiac insufficiency.
Hong-Yan Zhao et al.
BMC biotechnology, 18(1), 34-34 (2018-06-03)
Mesophilic alkaline serine proteases from various bacteria have been commercially applied in a range of industries owing to their high catalytic efficiency and wide substrate specificity. However, these proteases have an optimal catalytic temperature of approximately 50 °C, and their activity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service