Skip to Content
Merck
All Photos(1)

Key Documents

93711

Sigma-Aldrich

Atto 655

BioReagent, suitable for fluorescence, ≥85% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

Quality Level

Assay

≥85% (HPLC)

form

powder

manufacturer/tradename

ATTO-TEC GmbH

transmittance

254 nm
655 nm

fluorescence

λex 655 nm; λem 680 nm in 0.1 M phosphate pH 7.0

λ

in ethanol (with 0.1% trifluoroacetic acid)

UV absorption

λ: 652-658 nm Amax

suitability

suitable for fluorescence

storage temp.

−20°C

SMILES string

[S](=O)(=O)([O-])CC1CC(N(c2c1cc3c(c2)OC4=CC5=[N+](CCCC5=CC4=N3)CC)CCCC(=O)O)(C)C

InChI

1S/C27H33N3O6S/c1-4-29-9-5-7-17-11-20-24(13-22(17)29)36-25-14-23-19(12-21(25)28-20)18(16-37(33,34)35)15-27(2,3)30(23)10-6-8-26(31)32/h11-14,18H,4-10,15-16H2,1-3H3,(H-,31,32,33,34,35)

InChI key

FOYVTVSSAMSORJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Atto labels are designed for highest sensitivity applications. A unique combination of advantages makes them highly favorable tools for all kinds of labeling applications. Some of their properties make them specifically interesting for single molecule detection. Atto labels are based on rigid structures and do not show any cis-trans-isomerization, which lowers the brightness of signals and leads to environment dependency, e.g., spectral shifts by conjugation.
Atto 655 shows a molar extinction of 110,000 and QY of 30% in water (50% in ethanol). Decay time is 1.9 ns.

Other Notes

New red absorbing fluorescent dye with best signal-to-noise ratio and long fluorescence life-time. Useful as a biophysical probe for binding interactions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volker Buschmann et al.
Bioconjugate chemistry, 14(1), 195-204 (2003-01-16)
The spectroscopic characteristics (absorption, emission, and fluorescence lifetime) of 13 commercially available red-absorbing fluorescent dyes were studied under a variety of conditions. The dyes included in this study are Alexa647, ATTO655, ATTO680, Bodipy630/650, Cy5, Cy5.5, DiD, DY-630, DY-635, DY-640, DY-650
Eilon Sherman et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 12(3), 696-703 (2011-01-29)
Intramolecular dynamics in the denatured state of a protein are of importance for protein folding. Native-like contact formation within the ensemble of denatured conformations of a protein may guide its transformation towards the native conformation. The efficiency of folding is
Julie M G Rogers et al.
Langmuir : the ACS journal of surfaces and colloids, 27(7), 3815-3821 (2011-03-16)
The structure and function of the influenza A M2 proton channel have been the subject of intensive investigations in recent years because of their critical role in the life cycle of the influenza virus. Using a truncated version of the
Søren Preus et al.
Chembiochem : a European journal of chemical biology, 13(14), 1990-2001 (2012-09-01)
Förster resonance energy transfer (FRET) is a powerful tool for monitoring molecular distances and interactions at the nanoscale level. The strong dependence of transfer efficiency on probe separation makes FRET perfectly suited for "on/off" experiments. To use FRET to obtain
Achim Friedrich et al.
FEBS letters, 581(8), 1644-1648 (2007-04-03)
This article presents a new, highly sensitive method for the identification of single nucleotide polymorphisms (SNPs) in homogeneous solutions using fluorescently labeled hairpin-structured oligonucleotides (smart probes) and fluorescence single-molecule spectroscopy. While the hairpin probe is closed, fluorescence intensity is quenched

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service