Skip to Content
Merck
All Photos(1)

Key Documents

68338

Sigma-Aldrich

Hexylene glycol

BioUltra, ≥99.0% (GC)

Synonym(s):

(±)-2-Methyl-2,4-pentanediol, MPD

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH2C(CH3)2OH
CAS Number:
Molecular Weight:
118.17
Beilstein:
1098298
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.26

vapor density

4.1 (vs air)

vapor pressure

0.02 mmHg ( 20 °C)

product line

BioUltra

Assay

≥99.0% (GC)

form

liquid (or viscous liquid)

expl. lim.

7.4 %

impurities

insoluble matter, passes filter test

refractive index

n20/D 1.427 (lit.)
n20/D 1.428

pH

6-8 (25 °C, 1 M in H2O)

bp

197 °C (lit.)

mp

−40 °C (lit.)

solubility

H2O: 1 M at 20 °C, clear, colorless

density

0.925 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤1 mg/kg
As: ≤0.1 mg/kg
Ba: ≤1 mg/kg
Bi: ≤1 mg/kg
Ca: ≤5 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤20 mg/kg
Li: ≤1 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Mo: ≤1 mg/kg
Na: ≤20 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Sr: ≤1 mg/kg
Zn: ≤1 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: 0.01
λ: 280 nm Amax: 0.01

SMILES string

CC(O)CC(C)(C)O

InChI

1S/C6H14O2/c1-5(7)4-6(2,3)8/h5,7-8H,4H2,1-3H3

InChI key

SVTBMSDMJJWYQN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Used in the crystallisation of proteins

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kanchan Anand et al.
Acta crystallographica. Section D, Biological crystallography, 58(Pt 10 Pt 1), 1722-1728 (2002-09-28)
2-Methyl-2,4-pentanediol (MPD) is the most popular chemical additive used for crystallization of biological macromolecules. However, the mechanism of its action on proteins in aqueous solution is not well understood. We have carried out a systematic analysis of the conformation and
A.M. Brzozowski et al.
Journal of Applied Crystallography, 34, 97-97 (2001)
Catherine Michaux et al.
Journal of molecular biology, 375(5), 1477-1488 (2007-12-18)
Sodium dodecyl sulfate (SDS) is a highly effective and widely used protein denaturant. We show that certain amphipathic cosolvents such as 2-methyl-2,4-pentanediol (MPD) can protect proteins from SDS denaturation, and in several cases can refold proteins from the SDS-denatured state.
Jose Antonio Cuesta-Seijo et al.
Structure (London, England : 1993), 18(9), 1210-1219 (2010-09-10)
Enzymatic reactions involving bilayer lipids occur in an environment with strict physical and topological constraints. The integral membrane enzyme PagP transfers a palmitoyl group from a phospholipid to lipid A in order to assist Escherichia coli in evading host immune
Yoshikazu Tanaka et al.
Protein science : a publication of the Protein Society, 20(2), 448-456 (2011-02-01)
Staphylococcal α-hemolysin is expressed as a water-soluble monomeric protein and assembles on membranes to form a heptameric pore structure. The heptameric pore structure of α-hemolysin can be prepared from monomer in vitro only in the presence of deoxycholate detergent micelles

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service